Skip to Content
Merck
All Photos(1)

Key Documents

CLS3568

Corning® disposable GL45 solid cap for plastic spinner flasks

polyethylene solid cap for GL45 plastic spinner flask, disposable, sterile, 4/cs

Synonym(s):

Corning cap

Sign Into View Organizational & Contract Pricing

Select a Size

4 EA
$53.00

$53.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
4 EA
$53.00

About This Item

UNSPSC Code:
41121804
NACRES:
NB.17

$53.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

orange high-density polyethylene cap
polyethylene

sterility

sterile

packaging

pack of 1
case of 4

manufacturer/tradename

Corning 3568

color

orange

General description

These disposable high density polyethylene sterile solid caps are designed to fit the Corning® 500 mL, 1 L and 3 L Disposable Spinner Flasks (CLS3153, CLS3561and CLS3563). They also fit Corning disposable polystyrene and polycarbonate storage bottles and glass bottles with GL45 threads.
  • Individually double bagged
  • Features large knurls for ergonomic handling
  • Sterilized (10-6 SAL) by gamma radiation; autoclaving is not recommended

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service