CLS3567
Corning® disposable vent cap for GL45 plastic spinner flasks
vented cap for GL45 plastic spinner flasks, polyethylene, sterile, 4/cs
Synonym(s):
Corning® disposable vent cap for GL45 plastic spinner flasks, cell culture flasks, cell culture spinner flasks, scale up cell culture flasks
Select a Size
$57.00
Select a Size
About This Item
$57.00
Recommended Products
material
high-density polyethylene cap (vented)
polyethylene
sterility
sterile (γ-irradiated)
feature
autoclavable: no
disposable
thread finish: 45 GL
packaging
pack of 1
case of 4
manufacturer/tradename
Corning 3567
color
orange cap
Related Categories
General description
- Individually double bagged.
- Features large knurls for ergonomic handling.
- Sterilized (10-6 SAL) by gamma radiation; autoclaving is not recommend.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service