Key Documents
Z153958
Caps for Duran® laboratory bottles
GL 45 neck thread, red PBT (with PTFE seal)
Synonym(s):
caps bottles, glass bottles caps
Select a Size
10 EA
$187.00
$187.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
10 EA
$187.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
$187.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
red PBT (with PTFE seal)
manufacturer/tradename
DURAN®
Schott
parameter
-45-180 °C temp. stability
fitting
GL 45 neck thread
General description
The classic laboratory bottle GL threaded screw cap features the tried and tested cap plug sealing system ensuring a liquid tight seal. Available in four colours; blue, green, yellow and grey with matching pouring rings. Distinguishing media types is simplified and interchanging of screw caps and the risk of cross contamination is greatly reduced.
The red polybutylene terephthalate (PBT) GL cap and ETFE pouring rings offer greater thermal and chemical resistance than the equivalent polypropylene components. Screw cap seal has a PTFE faced cap liner for efficient sealing. ETFE pouring ring ensures drip free pouring.
Features and Benefits
  • High Temperature Screw Cap
  • High Performance
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)