A lip seal is lip or ring that is molded into the top of the cap. It fits against the interior wall of the tube or bottle preventing the contents from coming in contact with the seal or threads, thereby reducing the chances of sample contamination. See the link below for Duran pouring rings which are offered separately. There are several GL45 options:
https://www.sigmaaldrich.com/substance/pouringringsforduranlaboratorybottles1234598765
Z153923
Caps for Duran® laboratory bottles
GL 45 neck thread, blue polypropylene (with lip seal)
Synonym(s):
caps bottles, glass bottles caps
Select a Size
$48.20
Select a Size
About This Item
$48.20
Recommended Products
material
blue polypropylene (with lip seal)
manufacturer/tradename
DURAN®
Schott
parameter
-40-140 °C temp. stability
fitting
GL 45 neck thread
Related Categories
General description
Features and Benefits
- Cap with integral lip seal provides effective sealing for short term storage
- Pouring ring for drip-free pouring
- The colour helps prevent interchanging of cap mix-ups and reduces the risk of cross contamination
- Fully compatible with the GL 45 threads and therefore highly versatile
Legal Information
recommended
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Customers Also Viewed
Articles
A 4-step comprehensive ICP-MS workflow including sample homogenization, with standard addition calibration method, to determine heavy metals in Cannabis sativa.
-
It says caps with lip seal - is this the rings that go around the top of the bottle to make pouring easier? We ordered these caps and they did not come with those
1 answer-
Helpful?
-