Please see the link below to review the product datasheet which included the vector map and additional sequence details:
https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/262/320/crisprbul.pdf
To request the full sequence, we kindly ask you to navigate to the link https://www.sigmaaldrich.com/techservice, click on "Product Documentation" under the Products Section with all the required information so that a member of our team can reach out to you to assist further. Thank you.
CAS9GFPP
CMV-CAS9-2A-GFP Plasmid
Synonym(s):
Cas9 plasmid
Select a Size
Select a Size
About This Item
Recommended Products
recombinant
expressed in E. coli
packaging
vial of 50 μL
concentration
20 ng/μL in TE buffer; DNA (1μg of plasmid DNA)
Promoter
Promoter name: CMV
reporter gene
GFP
selection
kanamycin
shipped in
dry ice
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
CMV-CAS9-2A-GFP plasmid is suitable for functional genomics/target validation for:
- Creation of gene knockouts in multiple cell lines
- Complete knockout of genes not amenable to RNAi
- Creation of knock-in cell lines with promoters, fusion tags, or reporters integrated into endogenous genes
Components
Please note, product does not contain guideRNA sequence. This must be purchased separately through the Custom CRISPR product tab.
Principle
Other Notes
Typical transfection concentrations used in literature are in the ranges of >= 1.0 μg/μL and <= 5 μL of Cas9 plasmid combined with >= 1.0 μg/μL and <= 5 μL of U6-gRNA plasmids. (All dosages above assume 0.5 to 1 million cells nucleofected).
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
Learn about CRISPR Cas9, what it is and how it works. CRISPR is a new, affordable genome editing tool enabling access to genome editing for all.
-
Where can we find the sequence for this plasmid: CAS9GFPP
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service