Skip to Content
Merck
All Photos(1)

Key Documents

CRISPR17

Sigma-Aldrich

CRISPR Activator Human Oct4 Positive Control Plasmid

gRNA Only Expression Plasmid

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$443.00

$443.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 4 to 6 weeks.

Request a Bulk OrderRequest more information

Select a Size

Change View
1 EA
$443.00

About This Item

UNSPSC Code:
41106609
NACRES:
NE.02

$443.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 4 to 6 weeks.

Request a Bulk OrderRequest more information

Quality Level

form

liquid

packaging

vial of 50 μL

concentration

20 ng/μL in TE buffer; DNA (1μg of plasmid DNA)

application(s)

CRISPR

shipped in

dry ice

storage temp.

−20°C

Related Categories

General description

OCT4 is the key transcription factor (TF) that maintains pluripotency of embryonic stem cells and plays a critical role in reinstating cellular pluripotency. The expression of OCT4 is stringently silenced in differentiated cells. Activation of silenced OCT4 gene has become a hallmark event during epigenetic reprogramming into iPSCs. The positive control OCT4 gRNA design has been tested with the dCas9-p300 plasmid in HEK293 cells and shown to increase OCT4 mRNA transcript levels over three-fold compared to a negative control.

Application

Functional Genomics/Target Validation
  • Epigenetic Modification
  • Transcriptional Activation

Features and Benefits

Serves as an experimental control for the CRISPR dCas9-p300 activation system. A positive experiment will result in an increased level of mRNA via qPCR or TaqMan PCR assay.

Components

1 vial containing 1ug of U6-gRNA plasmid expressing a guide sequence to human OCT4 (Octamer-binding transcription factor-4 or POU5F1).

Linkage

To order U6-gRNA plasmids, please visit sigma.com/crisprs

Click here for more information on dcas9-p300 Gene Activator Expression Plasmid.

Other Notes

Must be used with dCas9-p300 or other dCas9 activator/repressor plasmids.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service