Skip to Content
Merck
All Photos(1)

Key Documents

CAS9P

Sigma-Aldrich

Cas9 plasmid

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$299.00

$299.00


Estimated to ship onMarch 19, 2025


Request a Bulk Order

Select a Size

Change View
1 EA
$299.00

About This Item

UNSPSC Code:
41106610
NACRES:
NA.51

$299.00


Estimated to ship onMarch 19, 2025


Request a Bulk Order

recombinant

expressed in E. coli

Quality Level

packaging

vial of 50 μL

concentration

20 ng/μL in TE buffer; DNA (1μg of plasmid DNA)

Promoter

Promoter name: CMV

selection

kanamycin

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The Cas9 expression plasmids use the CMV promoter for strong transient expression of Cas9. Alternate promoters can be substituted by replacement of CMV using MluI and NheI. Also, the Cas9 expression plasmids can be linearized using XbaI for T7-based mRNA production.

Application

Functional Genomics/Target Validation
  • Creation of gene knockouts in multiple cell lines
  • Complete knockout of genes not amenable to RNAi
  • Creation of knock-in cell lines with promoters, fusion tags or reporters integrated into endogenous genes

Components

1 vial containing 1ug of Cas9 plasmid.

Please note, product does not contain guideRNA sequence. This must be purchased separately through the Custom CRISPR product tab.

Principle

CRISPR/Cas systems are employed by bacteria and archaea as a defense against invading viruses and plasmids. Recently, the type II CRISPR/Cas system from the bacterium Streptococcus pyogenes has been engineered to function in eukaryotic systems using two molecular components: a single Cas9 protein and a non-coding guide RNA (gRNA). The Cas9 endonuclease can be programmed with a single gRNA, directing a DNA double-strand break (DSB) at a desired genomic location. Similar to DSBs induced by zinc finger nucleases (ZFNs), the cell then activates endogenous DNA repair processes, either non-homologous end joining (NHEJ) or homology-directed repair (HDR), to heal the targeted DSB.

Physical form

Sigma Cas9 plasmid DNA is supplied at concentrations of 20ng/ul in 50ul.

Preparation Note

Sigma CRISPR plasmid products are delivered as mini-prep aliquots, which may not be suitable for transfection into particular cell types. For best results, we advise maxi-prepping plasmids using endotoxin-free DNA purification kits prior to transfection.

Other Notes

Must be used in conjunction with a U6-gRNA plasmid in order to mediate a double strand break in the DNA.

Typical transfection concentrations used in literature are in the ranges of >= 1.0 ug/uL and <= 5 uL of Cas9 plasmid combined with >= 1.0 ug/uL and <= 5 uL of U6-gRNA plasmids. (All dosages above assume 0.5 to 1 million cells nucleofected)

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Wang et al.
Reproduction (Cambridge, England), 160(3), 353-365 (2020-06-11)
In this study, we investigated a gene-edited (Zp2MT/MT) rat model of infertility caused by the failure to express the zona pellucida glycoprotein 2 (ZP2) due to the significant reduction of mRNA amount. We examined the defects in the zona pellucida

Protocols

Learn about CRISPR Cas9, what it is and how it works. CRISPR is a new, affordable genome editing tool enabling access to genome editing for all.

Related Content

Sigma-Aldrich® Advanced Genomics is the leading provider of gene editing and silencing technologies including CRISPR, Cas9, synthetic guide RNA (sgRNA), and Zinc Finger Nuclease (ZFN).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service