Skip to Content
Merck
All Photos(1)

Key Documents

ESPCAS9RFP

Sigma-Aldrich

CMV-ESPCAS9-2A-RFP Plasmid

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$299.00

$299.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 4 to 6 weeks.

Request a Bulk Order

Select a Size

Change View
1 EA
$299.00

About This Item

UNSPSC Code:
41106609
NACRES:
NA.51

$299.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 4 to 6 weeks.

Request a Bulk Order

recombinant

expressed in E. coli

Quality Level

packaging

vial of 50 μL

concentration

20 ng/μL in TE buffer; DNA (1μg of plasmid DNA)

Promoter

Promoter name: CMV

reporter gene

RFP

selection

kanamycin

shipped in

dry ice

storage temp.

−20°C

General description

This product is an expression plasmid that utilizes the CMV promoter for strong transient expression of eSpCas9 and RFP linked by a 2A peptide (CMV-eSpCas9-2A-RFP) allowing for easy visualization of successful transfection. The eSpCas9 expression plasmid is one part of a two part CRISPR system with individual eSpCas9 and gRNA expression vectors.

To order gRNA in any format click here

Application

Functional Genomics/Target Validation
  • Creation of gene knockouts in multiple cell lines
  • Complete knockout of genes not amenable to RNAi
  • Creation of knock-in cell lines with promoters, fusion tags, or reporters integrated into endogenous genes

Features and Benefits

  • Enhanced specificity compared to wild type Cas9
  • Highly active
  • Ready to use purified plasmid DNA

Principle

CRISPR/Cas systems are employed by bacteria and archaea as a defense against invading viruses and plasmids. Recently, the type II CRISPR/Cas system from the bacterium Streptococcus pyogenes has been engineered to function in eukaryotic systems using two molecular components: a single Cas9 protein and a non-coding guide RNA (gRNA). The Cas9 endonuclease can be programmed with a single gRNA, directing a DNA double-strand break (DSB) at a desired genomic location. Similar to DSBs induced by zinc finger nucleases (ZFNs), the cell then activates endogenous DNA repair processes, either non-homologous end joining (NHEJ) or homology-directed repair (HDR), to heal the targeted DSB. Newly engineered eSpCas9 enables the efficient targeted gene editing of established CRISPR systems with the benefit of reduced off-target effects. Point mutations in the chromosome-binding motif of SpCas9, as described by Slaymaker, et al., provide higher on-target fidelity without loss of cleavage efficiency.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rationally engineered Cas9 nucleases with improved specificity.
Slaymaker, I.M., et al.
Science, 351, 84-88 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service