Key Documents
C2062
Bottle carrier, Nalgene®
Holds 1 x 4 L bottle
Synonym(s):
carrier bottle, nalgene carrier
Select a Size
1 EA
$290.00
$290.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onMay 31, 2025
Select a Size
Change View
1 EA
$290.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$290.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onMay 31, 2025
material
epoxy-coated handle
low-density polyethylene
feature
Holds 1 x 4 L bottle
packaging
pack of 1 ea
manufacturer/tradename
Nalgene® 6501-4000
General description
Handle locks cover while carrying.
Clear plastic cap protects bottle closure and allows identification of color-coded closure on bottles. Remove the cap with a twist and dispense without removing the bottle from the carrier.
Holds and protects standard acid and chemical bottles and permits pouring without removing the bottles.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Related Content
Packaging options for solvents include glass, PVC coated, Sure/Seal bottles, and aluminum cans, offering diverse dispensing solutions.