Skip to Content
Merck
All Photos(1)

Key Documents

A4486

Sigma-Aldrich

AMPK (α1/β1/γ3), active, His tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

HAMPKb, MGC17785, MGC33776, MGC57364, PRKAA1, PRKAB1, PRKAG3

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$1,990.00

$1,990.00


Estimated to ship onMay 07, 2025


Request a Bulk Order

Select a Size

Change View
10 μG
$1,990.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

$1,990.00


Estimated to ship onMay 07, 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

157-213 nmol/min·mg

mol wt

A1 subunit ~68 kDa
B1 subunit ~38 kDa
G3 subunit ~51 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

Biochem/physiol Actions

AMPK (A1/B1/G3) is a member of the AMPK family which are heterotrimeric proteins consisting of an α catalytic subunit, and non-catalytic β and γ subunits. AMPKs are an important energy-sensing enzyme group in the cells that monitor energy status particularly in response to stress. AMPKs regulate fatty acid and cholesterol synthesis by regulating the key rate-limiting enzymes acetyl-CoA carboxylase and 3-hydroxy-3-methylglutaryl-CoA reductase. The γ subunit is dominantly expressed in skeletal muscle where it may play a key role in the regulation of energy metabolism.

Physical form

Supplied in 50 mM Sodium Phosphate, pH 7.0, with 150 mM imidazole, 0.1 mM PMSF, 0.2 mM DTT, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P C Cheung et al.
The Biochemical journal, 346 Pt 3, 659-669 (2000-03-04)
The AMP-activated protein kinase (AMPK) cascade plays an important role in the regulation of energy homeostasis within the cell. AMPK is a heterotrimer composed of a catalytic subunit (alpha) and two regulatory subunits (beta and gamma). We have isolated and
B Viollet et al.
Biochemical Society transactions, 31(Pt 1), 216-219 (2003-01-28)
AMP-activated protein kinase (AMPK) is viewed as a fuel sensor for glucose and lipid metabolism. To understand better the physiological role of the catalytic AMPK subunit isoforms, we generated two knockout mouse models with the alpha1 (AMPK alpha 1(-/-)) and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service