Skip to Content
Merck
All Photos(1)

Key Documents

S5891

Sigma-Aldrich

Wheaton Coplin staining jars

5 slide jar with plastic screw cap

Synonym(s):

900520, Coplin Staining Jars, Staining Jars, Wheaton Staining Jars

Sign Into View Organizational & Contract Pricing

Select a Size

6 EA
$383.00

$383.00


Estimated to ship onApril 19, 2025



Select a Size

Change View
6 EA
$383.00

About This Item

UNSPSC Code:
41121800
NACRES:
MA.01

$383.00


Estimated to ship onApril 19, 2025


material

glass jar

feature

capacity 5 3 × 1 in. (76 × 25 mm) slide (vertically, ten slides back-to-back)
Wheaton staining ware is not recommended for any moist heat applications such as microwaves, autoclaves or water baths.

manufacturer/tradename

WHEATON® 900520

inside L × W × D

26 mm × 26 mm × 70 mm

Looking for similar products? Visit Product Comparison Guide

General description

Slides extend above the opening permitting them to be manipulated without the use of forceps.

Features and Benefits

• Manufactured from soda-lime glass• This unit holds ten 3” x 1” (75mmx 25mm) slides, back-to-back that extend above the opening so you can manipulate them without using forceps• Supplied with polypropylene screw cap (cat.no. 900522)• The screw cap is made of linerless white polypropylene to reduce solvent evaporation and spills during storage• Cap is not intended for use with slides in jar• The unit has a rectangular base and holds approximately 55mL• Approximate inside bottom dimensions (mm): 26 L x 26 W x 70 D

Legal Information

WHEATON is a registered trademark of Wheaton Industries, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Available Fluorescent in situ hybridization (FISH) procedures, reagents and equipment.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service