Skip to Content
Merck
All Photos(1)

Key Documents

BR472500

BRAND® glass staining trough, Schiefferdecker pattern

soda-lime glass

Synonym(s):

Microscopy, Staining, Staining Accessories

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$442.00

$442.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
10 EA
$442.00

About This Item

UNSPSC Code:
41122800
NACRES:
MA.01

$442.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

soda-lime glass

description

for use with transparent staining tray

manufacturer/tradename

BRAND 472500

L × W

76 mm × 26 mm , holds 10 slides

General description

The BRAND® Glass Staining Trough in the Schiefferdecker pattern is designed for the efficient staining of up to 10 slides, making it an essential tool for laboratory applications. Made from durable soda-lime glass, this trough features dimensions of 76 x 26 mm, providing a compact and effective solution for slide organization. The Schiefferdecker design allows for optimal spacing and accessibility, facilitating uniform staining while enhancing workflow efficiency. This practical and reliable staining trough is ideal for histology and microscopy procedures, making it a valuable addition to any laboratory setup.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service