Key Documents
S5392
Wheaton Coplin staining jars
Plastic screw cap only
Synonym(s):
240330
Select a Size
20 EA
$129.00
$129.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
20 EA
$129.00
About This Item
UNSPSC Code:
41121800
NACRES:
MA.01
$129.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
feature
Wheaton staining ware is not recommended for any moist heat applications such as microwaves, autoclaves or water baths.
manufacturer/tradename
WHEATON®
Features and Benefits
  • Aluminum foil faced pulp board liners can be used with organic solvents. Good resistance to hydrocarbons, oils, ketones and alcohols Not good for acids or alkalis.
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)