Skip to Content
Merck
All Photos(1)

Key Documents

60414

Sigma-Aldrich

Potassium nitrate

BioUltra, ≥99.5% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KNO3
CAS Number:
Molecular Weight:
101.10
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

BioUltra

Assay

≥99.5% (T)

impurities

insoluble matter, passes filter test

loss

≤0.1% loss on drying, 110 °C

pH

5.0-8.0 (25 °C, 1 M in H2O)

mp

334 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤10 mg/kg
iodate (IO3-): ≤5 mg/kg
nitrite (NO2-): ≤10 mg/kg
phosphate (PO43-): ≤5 mg/kg
sulfate (SO42-): ≤20 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤2 mg/kg
Li: ≤5 mg/kg
Mg: ≤50 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
NH4+: ≤10 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

absorption

cut-off at 328 nm in H2O at 1 M

SMILES string

[K+].[O-][N+]([O-])=O

InChI

1S/K.NO3/c;2-1(3)4/q+1;-1

InChI key

FGIUAXJPYTZDNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Substrate for the assay of nitrate reductase[1]; Component of media for testing bacterial nitrate reduction.

Pictograms

Flame over circle

Signal Word

Warning

Hazard Statements

Hazard Classifications

Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolation and characterization of nitrate reductase from Escherichia coli.
C H MacGregor
Methods in enzymology, 53, 347-355 (1978-01-01)
Rose A Devasia et al.
The Journal of antimicrobial chemotherapy, 63(6), 1173-1178 (2009-03-31)
The aim of this study was to assess the sensitivity, specificity and time to results of mycobacterial growth indicator tube (MGIT) 960, microscopic observation drug susceptibility (MODS) assay and nitrate reductase assay (NRA) compared with the gold standard agar proportion
Wolfgang R Engelsberger et al.
The Plant journal : for cell and molecular biology, 69(6), 978-995 (2011-11-09)
Nitrogen is an essential macronutrient for plant growth and development. Inorganic nitrogen and its assimilation products control various metabolic, physiological and developmental processes. Although the transcriptional responses induced by nitrogen have been extensively studied in the past, our work here
Mila Ivanova Luleva et al.
Sensors (Basel, Switzerland), 11(4), 4188-4206 (2011-12-14)
Displacement of soil particles caused by erosion influences soil condition and fertility. To date, the cesium 137 isotope ((137)Cs) technique is most commonly used for soil particle tracing. However when large areas are considered, the expensive soil sampling and analysis
Carlotta Pucci et al.
Soft matter, 10(48), 9657-9667 (2014-10-31)
The phase behavior of an ad hoc synthesized surfactant, sodium 8-hexadecylsulfate (8-SHS), and its mixtures with didecyldimethylammonium bromide (DiDAB) in water is reported. We dealt with dilute concentration regimes, at a total surfactant content of <30 mmol kg(-1) where vesicular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service