Skip to Content
Merck
All Photos(1)

Documents

1.05063

Supelco

Potassium nitrate

for analysis EMSURE® ISO,Reag. Ph Eur

Synonym(s):

Potassium nitrate, Nitric acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KNO3
CAS Number:
Molecular Weight:
101.10
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-818-8
NACRES:
NA.21

Agency

reag. ISO
reag. Ph. Eur.

Quality Level

product line

EMSURE®

Assay

≥99.0% (alkalimetric)

form

solid

potency

3015 mg/kg LD50, oral (Rat)

pH

5.0-7.5 (20 °C, 50 g/L in H2O)

mp

334 °C

bulk density

800 kg/m3

anion traces

chloride (Cl-): ≤0.001%
iodate (IO3-): ≤0.0005%
nitrite (NO2-): ≤0.001%
phosphate (PO43-): ≤0.0005%
sulfate (SO42-): ≤0.003%

cation traces

Ca: ≤0.001%
Cu: ≤0.0001%
Fe: ≤0.0003%
Mg: ≤0.0015%
NH4+: ≤0.001%
Na: ≤0.02%
Pb: ≤0.0001%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

InChI

1S/K.NO3/c;2-1(3)4/q+1;-1

InChI key

FGIUAXJPYTZDNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • The scientific rationale and development of an optimized dentifrice for the treatment of dentin hypersensitivity.: This clinical study details the development of a dentifrice containing potassium nitrate, aimed at treating dentin hypersensitivity, showcasing the compound′s application in dental health products (Tavss et al., 2004).


Analysis Note

Assay (alkalimetric): ≥ 99.0 %
pH-value (5 %; water): 5.0 - 7.5
Chloride (Cl): ≤ 0.001 %
Iodate (IO₃): ≤ 0.0005 %
Nitrite (NO₂): ≤ 0.001 %
Phosphate (PO₄): ≤ 0.0005 %
Sulfate (SO₄): ≤ 0.003 %
Heavy metals (as Pb): ≤ 0.0005 %
Ca (Calcium): ≤ 0.001 %
Cu (Copper): ≤ 0.0001 %
Fe (Iron): ≤ 0.0003 %
Mg (Magnesium): ≤ 0.0015 %
Na (Sodium): ≤ 0.02 %
NH₄ (Ammonium): ≤ 0.001 %
Pb (Lead): ≤ 0.0001 %
Corresponds to ISO,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circle

Signal Word

Warning

Hazard Statements

Hazard Classifications

Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service