Skip to Content
Merck
All Photos(1)

Key Documents

931950

Sigma-Aldrich

Sodium perchlorate

anhydrous, ≥99.9% trace metals basis

Synonym(s):

Sodium Perchlorate, Hyperchloric acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
ClNaO4
CAS Number:
Molecular Weight:
122.44
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23
grade:
anhydrous
battery grade
form:
powder
solubility:
H2O: 209 g/dL at 15 °C
Pricing and availability is not currently available.

grade

anhydrous
battery grade

Quality Level

Assay

≥99.9% trace metals basis

form

powder

impurities

≤1000 ppm (trace metals analysis)

pH

6.0-8.0 (25 °C, 5%, aq.sol.)

mp

482 °C

solubility

H2O: 209 g/dL at 15 °C

anion traces

chloride (Cl-): ≤30 ppm
sulfate (SO42-): ≤20 ppm

cation traces

Fe: ≤5 ppm
K: ≤500 ppm

application(s)

battery manufacturing

SMILES string

[Na+].[Cl](=O)(=O)(=O)[O-]

InChI

1S/ClHO4.Na/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1

InChI key

BAZAXWOYCMUHIX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Anhydrous sodium perchlorate is a white crystalline solid. It is hygroscopic and absorbs water to form its monohydrate. Anhydrous sodium perchlorate is highly soluble in water, and soluble in a range of polar organic solvents such as methanol, ethanol, acetone, carbonates (including ethylene carbonate, dimethyl carbonate, propylene carbonate, and diethyl carbonate), and ethers (including dimethoxyethane, tetrahydrofuran, and triethylene glycol dimethyl ether). It is insoluble in benzene, chloroform, and toluene.

Application

The major application of anhydrous sodium perchlorate is as an electrolyte in sodium-ion batteries. It is popular because of its solubility in ethers and carbonates, its wide electrochemical stability window (e.g. from 0 to 5 V vs Na+/Na in propylene carbonate, triglyme, or diethylcarbonate)[1], and its compatibility with a wide range of materials. It has been used in batteries with hard-carbon anodes[2], mesoporous carbon anodes[3], sodium cobalt oxide cathodes (NaxCoO2)[4], sodium vanadium oxide cathodes (NaxVO2)[5], titanium dioxide cathodes[6], and emerging materials like high-entropy layered oxide cathodes[7].

Packaging

10 g in glass bottle
25 g in glass bottle

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 1 - STOT RE 2

Target Organs

Thyroid

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In search of an optimized electrolyte for Na-ion batteries
Ponrouch, A., et al.
Energy & Environmental Science, 5, 8572-8583 (2012)
Electrochemical Na Insertion and Solid Electrolyte Interphase for Hard-Carbon Electrodes and Application to Na-Ion Batteries.
Komaba, S., et al.
Advances in Functional Materials, 21, 3859-3867 (2011)
Gianluca Longoni et al.
Nano letters, 17(2), 992-1000 (2016-12-28)
Rechargeable sodium-ion batteries are becoming a viable alternative to lithium-based technology in energy storage strategies, due to the wide abundance of sodium raw material. In the past decade, this has generated a boom of research interest in such systems. Notwithstanding
Chenglong Zhao et al.
Angewandte Chemie (International ed. in English), 59(1), 264-269 (2019-10-18)
Material innovation on high-performance Na-ion cathodes and the corresponding understanding of structural chemistry still remain a challenge. Herein, we report a new concept of high-entropy strategy to design layered oxide cathodes for Na-ion batteries. An example of layered O3-type NaNi0.12
Jia Ding et al.
ACS nano, 7(12), 11004-11015 (2013-11-07)
We demonstrate that peat moss, a wild plant that covers 3% of the earth's surface, serves as an ideal precursor to create sodium ion battery (NIB) anodes with some of the most attractive electrochemical properties ever reported for carbonaceous materials.

Articles

Discover the role of electrolytes in sodium-ion batteries, to enhance performance, safety, and sustainability in energy storage solutions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service