Skip to Content
Merck
All Photos(2)

Key Documents

208051

Sigma-Aldrich

Sodium hexafluorophosphate

98%

Synonym(s):

Sodium hexafluorophosphate(v)

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$66.20
50 G
$297.00
250 G
$1,010.00

$66.20


Estimated to ship onMarch 28, 2025


Request a Bulk Order

Select a Size

Change View
10 G
$66.20
50 G
$297.00
250 G
$1,010.00

About This Item

Linear Formula:
NaPF6
CAS Number:
Molecular Weight:
167.95
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
98%
form:
powder and chunks
solubility:
H2O: slightly soluble(lit.)

$66.20


Estimated to ship onMarch 28, 2025


Request a Bulk Order

Quality Level

Assay

98%

form

powder and chunks

mp

>200 °C (lit.)

solubility

H2O: slightly soluble(lit.)

density

2.369 g/mL at 25 °C (lit.)

SMILES string

[Na+].F[P-](F)(F)(F)(F)F

InChI

1S/F6P.Na/c1-7(2,3,4,5)6;/q-1;+1

InChI key

KMADQUOCJBLXRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sodium hexafluorophosphate (SHFP) can be used as a precursor to prepare electrolytes for sodium-ion batteries.

SHFP bound amberlite resin can be used as a catalyst to synthesize benzimidazoles and quinoxalines.

Itcan also be used as an ion-pairing agent for the determination of inorganicspecies in saline solutions.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Zhao et al.
Small (Weinheim an der Bergstrasse, Germany), 14(20), e1703717-e1703717 (2018-04-17)
Na metal anode attracts increasing attention as a promising candidate for Na metal batteries (NMBs) due to the high specific capacity and low potential. However, similar to issues faced with the use of Li metal anode, crucial problems for metallic
Development of an in situ solvent formation microextraction and preconcentration method based on ionic liquids for the determination of trace cobalt (II) in water samples by flame atomic absorption spectrometry
Mohammed Reza Jamali, et al.
Arabian Journal of Chemistry (2012)
Xiang Huang et al.
The Analyst, 140(17), 6015-6024 (2015-07-21)
An efficient aptasensor was developed in which graphene oxide (GO) was employed as an indicator for both electrochemical impedance spectroscopy and electrochemiluminescence (ECL) signal generation. The aptasensor was fabricated by self-assembling the ECL probe of a thiolated adenosine triphosphate binding

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service