Skip to Content
Merck
All Photos(1)

Key Documents

PFS11

Supelco

SpheriCal Mix

Peptide Low, suitable for MALDI-MS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

product line

SpheriCal

form

powder

shelf life

expiry date on the label

mol wt

Mr 700-1500 Da

composition

9-nitroanthracene, >90%
sodium trifluoroacetate, <10%

technique(s)

MALDI-MS: suitable

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

calibrants PFS1A : PFS1B : PFS1C : PFS1D = 1:1:1:1
Matrix/counter ion: 9-Nitroanthracene/Na+

Application

For mass calibration in mass spectrometry

Packaging

Quantity: 5 mg/vial

Legal Information

SpheriCal is a trademark of Polymer Factory Sweden AB

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Grayson research labs have focused on the synthesis and characterization of well-defined polymers. One particular theme of research has involved the optimization of the mass spectral analysis of polymers using MALDI-TOF MS and associated calibration methodologies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service