Skip to Content
Merck
All Photos(1)

Key Documents

92817

Supelco

9-Aminoacridine

matrix substance for MALDI-MS, ≥99.5% (HPLC)

Synonym(s):

Aminacrine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10N2
CAS Number:
Molecular Weight:
194.23
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

grade

matrix substance for MALDI-MS

Quality Level

Assay

≥99.5% (HPLC)

form

powder

analyte functional class(es)

metabolites

analyte chemical class(es)

lipids

technique(s)

MALDI-MS: suitable

impurities

≤0.05% sulfated ash

loss

≤10% loss on drying

mp

238-240 °C

cation traces

Ba: ≤5 mg/kg
Ca: ≤200 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Fe: ≤10 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

Nc1c2ccccc2nc3ccccc13

InChI

1S/C13H10N2/c14-13-9-5-1-3-7-11(9)15-12-8-4-2-6-10(12)13/h1-8H,(H2,14,15)

InChI key

XJGFWWJLMVZSIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

9-Aminoacridine, an aminoacridine dye which acts as a potent mutagen in viruses and bacteria.

Application

A MALDI matrix exhibiting low background and good sensitivity in the analysis of low molecular weight anions, such as metabolites, in negative mode

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mass spectrometric method for analyzing metabolites in yeast with single cell sensitivity.
Andrea Amantonico et al.
Angewandte Chemie (International ed. in English), 47(29), 5382-5385 (2008-06-11)
9-Aminoacridine as a matrix for negative mode matrix-assisted laser desorption/ionization
Vermillion-Salsbury, R. L. and Hercules, D. M.
Rapid Communications in Mass Spectrometry, 16, 1575-1581 (2002)
Mutagen-nucleic acid intercalative binding: structure of a 9-aminoacridine: 5-iodocytidylyl (3'-5') guanosine crystalline complex.
Sakore, T. D., et al.
Proceedings of the National Academy of Sciences of the USA, 74.1, 188-192 (1977)
Fred Fack et al.
EMBO molecular medicine, 9(12), 1681-1695 (2017-10-22)
Heterozygous mutations in NADP-dependent isocitrate dehydrogenases (IDH) define the large majority of diffuse gliomas and are associated with hypermethylation of DNA and chromatin. The metabolic dysregulations imposed by these mutations, whether dependent or not on the oncometabolite D-2-hydroxyglutarate (D2HG), are
Daniel J Karst et al.
Biotechnology progress, 33(4), 879-890 (2016-12-21)
Perfusion cell culture processes allow the steady-state culture of mammalian cells at high viable cell density, which is beneficial for overall product yields and homogeneity of product quality in the manufacturing of therapeutic proteins. In this study, the extent of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service