Skip to Content
Merck
All Photos(1)

Key Documents

932183

Sigma-Aldrich

Hexaammineruthenium(III) chloride ChemBeads

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H18Cl3N6Ru
CAS Number:
Molecular Weight:
309.61
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.22
Pricing and availability is not currently available.

form

solid

Quality Level

storage temp.

2-8°C

SMILES string

[Ru+3].[Cl-].[Cl-].[Cl-].N.N.N.N.N.N

InChI

1S/3ClH.6H3N.Ru/h3*1H;6*1H3;/q;;;;;;;;;+3/p-3

InChI key

GBDZMMXUOBAJMN-UHFFFAOYSA-K

General description

Hexaammineruthenium III/II (HexRu(III)|HexRu(II)) couple is a commonly used electrochemical redox couple due to its chemical and electrochemical reversibility.

Application

Hexaammineruthenium(III) chloride can be used to synthesize chloropentaammineruthenium(III) chloride.

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

Learn more about ChemBeads products

Product is also available as neat precatalyst 262005

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Versatile Methods to Dispense Submilligram Quantities of Solids Using Chemical-Coated Beads for High-Throughput Experimentation
Martin, et al.
Organic Process Research & Development, 23, 1900?1907-1900?1907 (2019)
Wei Lu et al.
Biosensors & bioelectronics, 90, 258-263 (2016-12-05)
Rolling circle amplification (RCA) is an isothermal amplification technique with high efficiency and perfect accuracy for nucleic acids detection. However, RCA technique suffers the limitation to detect short DNA or RNA molecules. For long nucleic acid molecules, enzymatic restriction as
Misha Liu et al.
Analytical and bioanalytical chemistry, 409(29), 6821-6829 (2017-10-01)
Detection of DNA damage caused by ∙OH or radiation has led to rapidly growing interest in the fields of drug development, biochemistry, clinic diagnostics, and environmental evaluation. Electrochemical methods have been applied for DNA damage detection because of their fast
Rui Campos et al.
Langmuir : the ACS journal of surfaces and colloids, 34(29), 8472-8479 (2018-06-26)
Electrical properties of DNA critically depend on the way DNA molecules are integrated within the electronics, particularly on DNA-electrode immobilization strategies. Here, we show that the rate of electron transport in DNA duplexes spacer-free tethered to gold via the adenosine
A Erofeev et al.
Scientific reports, 8(1), 7462-7462 (2018-05-12)
Iron oxide nanoparticles have attracted a great deal of research interest and have been widely used in bioscience and clinical research including as contrast agents for magnetic resonance imaging, hyperthermia and magnetic field assisted radionuclide therapy. It is therefore important

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service