Skip to Content
Merck
All Photos(3)

Key Documents

919772

Sigma-Aldrich

Tris(dibenzylideneacetone)dipalladium(0) ChemBeads

Synonym(s):

Pd2dba3, Pd2(dba)3

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$75.60
1 G
$194.00

$75.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$75.60
1 G
$194.00

About This Item

Linear Formula:
(C6H5CH=CHCOCH=CHC6H5)3Pd2
CAS Number:
Molecular Weight:
915.72
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

$75.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

composition

~ 5 wt.% loading of catalyst

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

storage temp.

2-8°C

SMILES string

O=C(/C=C/c1ccccc1)/C=C/c2ccccc2.O=C(/C=C/c3ccccc3)/C=C/c4ccccc4.O=C(/C=C/c5ccccc5)/C=C/c6ccccc6.[Pd].[Pd]

InChI

1S/3C17H14O.2Pd/c3*18-17(13-11-15-7-3-1-4-8-15)14-12-16-9-5-2-6-10-16;;/h3*1-14H;;/b3*13-11+,14-12+;;

InChI key

CYPYTURSJDMMMP-WVCUSYJESA-N

Application

Application Guide for Palladium Catalyzed Cross-Coupling Reactions

Reactant involved in:
  • Synthesis of nanosized palladium phosphides upon interaction with white phosphorous
  • Preparation of palladium triphenylphosphine carbonyl cluster complexes
  • Precursor for synthesis of functionalized multiwalled carbon nanotube-palladium complexes used as catalysts for Heck coupling reactions
  • Selective carbon-sulfur bond formation via addition of S-S and S-H bonds to alkynes

Catalyst for:
  • Suzuki cross-coupling reactions
  • PCN- and PCS-pincer palladium complex catalyzed tandem allylation
Umicore Precatalysts for Asymmetric and Cross-Coupling Catalysis

Catalyst for C-C and C-N bond formation especially Suzuki, Heck coupling reaction

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.
Learn more about ChemBeads products
For larger scale uses, product also available in powdered form (328774)

Legal Information

Tris(dibenzylideneacetone)dipalladium(0) is Product of Umicore

Additional information available at www.pmc.umicore.com

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B Inhalation - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Versatile Methods to Dispense Sub-Milligram Quantities of Solids using Chemical Coated Beads for High-Throughput Experimentation.
Martin C M ,et al
Organic Process Research & Development, 23(9), 1900-1907 (2019)
George R Kracke et al.
ChemMedChem, 10(1), 62-67 (2014-11-26)
Clinically there is a need for local anesthetics with a greater specificity of action on target cells and longer duration. We have synthesized a series of local anesthetic derivatives we call boronicaines in which the aromatic phenyl ring of lidocaine
Michael J Ardolino et al.
Tetrahedron, 71(37), 6409-6413 (2015-08-19)
While Pd-catalyzed allyl-allyl cross-couplings in the presence of small-bite-angle bidentate ligands reliably furnish the branched regioisomer with high levels of selectivity, cross-couplings in the presence of large-bite-angle bidentate ligands give varying, often unpredictable, levels of selectivity. In a combined computational
Noah P Tu et al.
Angewandte Chemie (International ed. in English), 58(24), 7987-7991 (2019-03-21)
Technologies that enable rapid screening of diverse reaction conditions are of critical importance to methodology development and reaction optimization, especially when molecules of high complexity and scarcity are involved. The lack of a general solid dispensing method for chemical reagents
Ana L Aguirre et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 27(51), 12981-12986 (2021-07-08)
High-throughput experimentation (HTE) methods are central to modern medicinal chemistry. While many HTE approaches to C-N and Csp2 -Csp2 bonds are available, options for Csp2 -Csp3 bonds are limited. We report here how the adaptation of nickel-catalyzed cross-electrophile coupling of

Articles

ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service