Skip to Content
Merck
All Photos(1)

Key Documents

P5244

Sigma-Aldrich

Phosphate buffer solution

0.1 M

Synonym(s):

PBS

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$71.40

$71.40

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$71.40

About This Item

UNSPSC Code:
12161700
eCl@ss:
32129211
NACRES:
NA.25

$71.40

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

sterility

autoclaved

Quality Level

form

liquid

does not contain

preservatives

concentration

0.1 M

pH

7.5 (25 °C)

application(s)

diagnostic assay manufacturing

storage temp.

2-8°C

Application

Phosphate buffer solution has been used:
  • in the inhibition assay of α-glucosidase activity[1]
  • for measuring calcium deposition[2]
  • in trypsin grafting[3]

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diverse effect of BMP-2 homodimer on mesenchymal progenitors of different origin
Hrubi E, et al.
Human Cell : Official Journal of Human Cell Research Society, 31(2), 139-148 (2018)
alpha-glucosidase inhibitor activity of Terminalia species
Anam K, et al.
Int. J. Pharmacol., 5(4), 277-280 (2009)
Enhancing surface activity in silicon microreactors: use of black silicon and alumina as catalyst supports for chemical and biological applications.
Roumanie M, et al.
Chemical Engineering Journal, 135, S317-S326 (2008)
Stefan de Vries et al.
Tissue engineering. Part A, 25(11-12), 830-841 (2018-05-10)
Notochordal cells (NCs) reside in the core of the healthy disc and produce soluble factors that can stimulate nucleus pulposus cells (NPCs). These NC-derived factors may be applied in intervertebral disc regeneration for treatment of low-back pain. However, identification of
S A H de Vries et al.
Scientific reports, 8(1), 8875-8875 (2018-06-13)
Notochordal cell derived matrix (NCM) can induce regenerative effects on nucleus pulposus cells and may exert such effects on chondrocytes as well. Furthermore, when dissolved at low concentrations, NCM forms a viscous fluid with potential lubricating properties. Therefore, this study

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What is the formulation of this phosphate buffer solution please?

    1 answer
    1. This product is prepared from potassium phosphate monobasic. Sodium hydroxide is used to adjust the pH, but no additional sodium chloride has been added.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service