Skip to Content
Merck
All Photos(1)

Key Documents

75405

Millipore

Orange-serum Agar

suitable for microbiology, NutriSelect® Basic

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$221.00

$221.00


Estimated to ship onApril 08, 2025


Request a Bulk Order

Select a Size

Change View
500 G
$221.00

About This Item

UNSPSC Code:
41171607
NACRES:
NA.74

$221.00


Estimated to ship onApril 08, 2025


Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 17 g/L
casein peptone, 10 g/L
dipotassium hydrogen phosphate, 3 g/L
D(+)-glucose, 4 g/L
orange extract, 5 g/L
yeast extract, 3 g/L

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

final pH

5.5±0.2 (25 °C)

application(s)

bioburden testing
environmental
food and beverages

microbiology

suitability

selective by low pH for Aspergillus spp.
selective by low pH for Bacillus spp.
selective by low pH for Candida spp.
selective by low pH for Lactobacillus spp.
selective by low pH for Penicillium spp.
selective by low pH for Saccharomyces spp.
selective by low pH for aciduric bacteria
selective by low pH for bacteria (General Media)
selective by low pH for molds (General Media)
selective by low pH for yeasts (General Media)

Application

Orange-serum Agar can be used for the isolation, cultivation and enumeration of acid-tolerant spoilage microorganisms in fruit juice and fruit juice concentrates, in particular from citrus fruit, according to Hays, Troy and Beisel.
The prepared agar may be turbid after autoclaving due to orange extract.
Orange-serum Agar has been used to prepare modified orange serum (MOS) as a medium for Hymenoscyphus fraxineus.[1]

Preparation Note

Dissolve 42 g in 1 litre distilled water. Autoclave gently at 115°C for 15 minutes taking care not to overheat. The prepared agar may be turbid after autoclaving due to orange extract.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ADA, a fast-growth medium for Hymenoscyphus fraxineus
L. Botella
Forest Pathology, 46 (2016)
D.I. Murdock et al.
Food Technology, 6, 181-181 (1952)
G.L. Hays et al.
Food Technology, 6, 386-386 (1952)

Articles

Cost-effective molecular biology method for lactobacilli detection is presented as an alternative to PCR-based methods.

Yeast is one of the most important microorganisms known and utilised by mankind. Ancient Middle Eastern civilisations used the organism to bake bread and to produce mead, beer and wine.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service