Skip to Content
Merck
All Photos(1)

Key Documents

43479

Millipore

MGYP Agar with Copper

NutriSelect® Plus, suitable for microbiology

Synonym(s):

Copper Sulphate Agar, MYGP Agar, Malt extract Yeast extract Glucose Peptone Agar, Mould Yeast Agar with Copper

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$204.00

$204.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

Select a Size

Change View
500 G
$204.00

About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

$204.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 20.0 g/L
cupric sulfate, 0.4 g/L
dextrose, 10.0 g/L
gelatin peptone, 5.0 g/L
malt extract, 3.0 g/L
yeast extract, 3.0 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

6.2±0.2 (25 °C)

application(s)

food and beverages
microbiology

suitability

Candida spp.
Pichia spp.
Saccharomyces spp.
Zygosaccharomyces spp.

Application

MGYP Agar with Copper is a selective medium recommended for isolation and cultivation of wild yeasts in the brewing industry.

Preparation Note

Suspend 41.4 grams in 1000 ml distilled water. Heat to boiling to dissolve the medium completely. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes. Mix well and pour into sterile Petri plates.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service