Skip to Content
Merck
All Photos(1)

Key Documents

95765

Millipore

YGC Agar

suitable for microbiology, NutriSelect® Basic

Synonym(s):

Yeast Extract Glucose Chloramphenicol Agar FIL-IDF

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$237.00

$237.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
500 G
$237.00

About This Item

UNSPSC Code:
41171609
NACRES:
NA.74

$237.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 14.9 g/L
chloramphenicol, 0.1 g/L
D(+)-glucose, 20 g/L
yeast extract, 5 g/L

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

final pH

6.6±0.2 (25 °C)

application(s)

bioburden testing
environmental
food and beverages

microbiology

suitability

selective for Aspergillus spp.
selective for Candida spp.
selective for Penicillium spp.
selective for Pichia spp.
selective for Saccharomyces spp.
selective for Zygosaccharomyces spp.
selective for molds (General Media)
selective for yeasts (General Media)

Application

Selective agar for the enumeration and isolation of yeasts and molds in dairy products.

Preparation Note

Dissolve 40 g in 1 litre distilled water. Sterilize by autoclaving at 121°C for 15 minutes.

Other Notes

Comparison of media for the detection of yeasts and molds in dairy products[1]

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

G. Engel
Milchwiss., 37, 727-727 (1982)
Renata Maria Sumalan et al.
Molecules (Basel, Switzerland), 24(11) (2019-05-31)
Essential oils (EOs) are a natural source of active compounds with antifungal, antimycotoxigenic, and herbicidal potential, and have been successfully used in organic agriculture, instead of chemical compounds obtained by synthesis, due to their high bioactivity and the absence of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service