Skip to Content
Merck
All Photos(1)

Documents

70133

Millipore

Blood Agar (Base)

NutriSelect® Plus, suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171605
NACRES:
NA.74

sterility

non-sterile

Quality Level

product line

BioChemika

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15 g/L
meat extract, 10 g/L
peptone, 10 g/L
sodium chloride, 5 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | specific enzyme detection: suitable
microbiological culture: suitable

final pH

7.3±0.2 (25 °C)

application(s)

agriculture
clinical testing
environmental
food and beverages

microbiology

suitability

Aeromonas spp.
Haemophilus spp.
Pneumococcus spp.
nonselective and differential for Bacillus cereus
nonselective and differential for Bacillus spp.
nonselective and differential for Bordetella spp.
nonselective and differential for Candida spp.
nonselective and differential for Clostridium spp.
nonselective and differential for Corynebacterium spp.
nonselective and differential for Enterococcus spp.
nonselective and differential for Escherichia coli
nonselective and differential for Listeria spp.
nonselective and differential for Neisseria spp.
nonselective and differential for Pseudomonas spp.
nonselective and differential for Salmonella spp.
nonselective and differential for Shigella spp.
nonselective and differential for Staphylococcus spp.
nonselective and differential for Streptococcus spp.
nonselective and differential for bacteria (General Media)
nonselective and differential for coliforms

Application

A non-selective medium for the isolation and cultivation of many pathogenic and non-pathogenic microorganisms.

Preparation Note

Suspend 40 g in 1 litre of distilled water. Bring to the boil to dissolve completely. Sterilize by autoclaving at 121°C for 15 minutes. For blood agar, cool to 45-50°C and add aseptically 6% of sterile defibrinated blood.

Other Notes

Isolation of Aeromonas spp. from feces

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J Robinson et al.
Journal of medical microbiology, 18(3), 405-411 (1984-12-01)
Five solid media were evaluated for isolation of Aeromonas spp. from faeces: desoxycholate citrate agar (DCA), MacConkey's agar (MAC), xylose-desoxycholate-citrate agar (XDCA), Rogol's medium (ROG), which contained ampicillin 20 mg/L and p-nitrophenyl-glycerine 25 mg/L as inhibitors, and blood agar (BA)
Amanda Croasdell et al.
Journal of immunology (Baltimore, Md. : 1950), 196(6), 2742-2752 (2016-02-05)
Nontypeable Haemophilus influenzae (NTHi) is a Gram-negative, opportunistic pathogen that frequently causes ear infections, bronchitis, pneumonia, and exacerbations in patients with underlying inflammatory diseases, such as chronic obstructive pulmonary disease. In mice, NTHi is rapidly cleared, but a strong inflammatory
Blood media for the cultivation of Mycobacterium tuberculosis.
M S TARSHIS et al.
American journal of clinical pathology, 21(2), 101-113 (1951-02-01)
Slim Mzoughi et al.
Nature communications, 11(1), 3520-3520 (2020-07-16)
PRDM (PRDI-BF1 and RIZ homology domain containing) family members are sequence-specific transcriptional regulators involved in cell identity and fate determination, often dysregulated in cancer. The PRDM15 gene is of particular interest, given its low expression in adult tissues and its
Peter R Wilker et al.
Nature immunology, 9(6), 603-612 (2008-04-29)
Calcineurin is required for B cell receptor (BCR)-induced proliferation of mature B cells. Paradoxically, loss of NFAT transcription factors, themselves calcineurin targets, induces hyperactivity, which suggests that calcineurin targets other than NFAT are required for BCR-induced proliferation. Here we demonstrate

Articles

Clostridia are relatively large, gram-positive, rod-shaped bacteria that can undergo only anaerobic metabolism.

Chromogenic media enable the selective detection of S. aureus, which produce bluish-green colonies that are clearly differentiated from other species.

An article concerning the detection, identification, differentiation, and cultivation of Pseudomonas species.

Streptococci- Overview of Detection, Identification, Differentiation and Cultivation Techniques

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service