Skip to Content
Merck
All Photos(1)

Documents

03-0070

Sigma-Aldrich

Sodium 5,5-diethylbarbiturate

SAJ special grade, ≥98.0%

Synonym(s):

5,5-Diethylbarbituric acid sodium salt, Barbitone, Sodium barbital, Veronal sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11N2NaO3
CAS Number:
Molecular Weight:
206.17
Beilstein:
3921202
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

SAJ special grade

Assay

≥98.0%

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

availability

available only in Japan

SMILES string

[Na+].CCC1(CC)C(=O)NC([O-])=NC1=O

InChI

1S/C8H12N2O3.Na/c1-3-8(4-2)5(11)9-7(13)10-6(8)12;/h3-4H2,1-2H3,(H2,9,10,11,12,13);/q;+1/p-1

InChI key

RGHFKWPGWBFQLN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Sedative/hypnotic

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 2 - STOT SE 3

Target Organs

Nervous system, Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service