Skip to Content
Merck
All Photos(1)

Key Documents

SML2364

Sigma-Aldrich

4-Androstenedione

≥98% (HPLC)

Synonym(s):

4-Androstene-3,17-dione, (8R,9S,10R,13S,14S)-10,13-dimethyl-1,6,7,8,9,10,11,12,13,14,15,16-dodecahydro-3H-cyclopenta[a]phenanthrene-3,17(2H)-dione, 17-Ketotestosterone, 3,17-Dioxoandrost-4-ene, Androst-4-ene-3,17-dione, Androstenedione, Fecundin, NSC 9563, SKF 2170

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$74.00
500 MG
$149.00

$74.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$74.00
500 MG
$149.00

About This Item

Empirical Formula (Hill Notation):
C19H26O2
CAS Number:
Molecular Weight:
286.41
Beilstein:
2059239
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$74.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

drug control

USDEA Schedule III

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@]4(C)C(=O)CC[C@@]24[H]

InChI

1S/C19H26O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h11,14-16H,3-10H2,1-2H3/t14-,15-,16-,18-,19-/m0/s1

InChI key

AEMFNILZOJDQLW-QAGGRKNESA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

4-Androstenedione, commonly known as Androstenedione, is an endogenous androgen steroid hormone. 4-Androstenedione functions as an intermediate in the biosynthesis of testosterone, estradiol and estrone. Recent studies indicate that 4-Androstenedione is involved in aromatase inhibitors induce painful musculoskeletal symptoms (AIMSS) by activation of TRPA1.
Testosterone precursor and metabolite with androgenic activity.
Testosterone precursor and metabolite with androgenic activity; may increase the risk of prostate or pancreatic cancer.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesco De Logu et al.
Cancer research, 76(23), 7024-7035 (2016-10-21)
Aromatase inhibitors (AI) induce painful musculoskeletal symptoms (AIMSS), which are dependent upon the pain transducing receptor TRPA1. However, as the AI concentrations required to engage TRPA1 in mice are higher than those found in the plasma of patients, we hypothesized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service