Skip to Content
Merck
All Photos(1)

Key Documents

10108987001

Roche

Pyrophosphatase, inorganic (PPase)

from yeast

Synonym(s):

inorganic pyrophosphatase

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352204
Pricing and availability is not currently available.

biological source

yeast

Quality Level

form

suspension

specific activity

~200 units/mg protein (At 25 °C with inorganic pyrophosphate as the substrate.)

packaging

pkg of 1 mL (1 mg)

manufacturer/tradename

Roche

storage temp.

2-8°C

General description

Pyrophosphate phosphohydrolase
The enzyme pyrophosphatase is ubiquitously distributed in a number of organisms.[1]

Specificity

Specific activity: Approximately 200 U/mg at +25°C with inorganic pyrophosphate as the substrate.

Application

Inorganic pyrophosphatase has been used in enzyme assays and enzyme inhibition studies.[2]
Pyrophosphatase, inorganic (PPase) has been used in :
  • Enzymatic nitric oxide (NO) detection system[3]
  • In vitro transcription [4][5]
  • Multiply-primed rolling circle amplification[6]

Biochem/physiol Actions

Inorganic pyrophosphatase catalyzes the hydrolysis of inorganic pyrophosphate to orthophosphate.[1]
Mg ions are necessary for catalytic activity of crystalline pyrophosphatase.[1]

Quality

Contaminants: <0.01% ATPase and phosphatases, each (at pH 7 and 4-nitrophenyl phosphate as the substrate).

Physical form

Suspension in 3.2 M ammonium sulfate solution, pH approximately 6

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael A Kron et al.
Protein science : a publication of the Protein Society, 17(6), 983-989 (2008-04-16)
Diagnosis of eukaryotic parasitic infection using antibody-based tests such as ELISAs (enzyme-linked immunosorbent assays) is often problematic because of the need to differentiate between homologous host and pathogen proteins and to ensure that antibodies raised against a peptide will also
Single prokaryotic cell isolation and total transcript amplification protocol for transcriptomic analysis.
Kang Y, et al.
Nature Protocols, 10(7), 974-974 (2015)
Tracking the m7G-cap during translation initiation by crosslinking methods.
Gross L, et al.
Methods, 137, 3-10 (2018)
2'-O-methylation of the mRNA cap protects RNAs from decapping and degradation by DXO.
Picard-Jean F, et al.
PLoS ONE, 13(3), e0193804-e0193804 (2018)
Assimilatory sulfate reduction in the marine methanogen Methanothermococcus thermolithotrophicus.
Jespersen, et al.
Nature microbiology, 8, 1227-1239 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service