Skip to Content
Merck
All Photos(1)

Key Documents

11277057001

Roche

Ribonucleoside Triphosphate Set

pkg of 4 × 200 μL (4 x 20 μmol; 100 mM, each), suitable for DNA sequencing, >98% (HPLC)

Synonym(s):

atp, ctp, gtp, set of atp, ctp, gtp, utp, utp

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116100

description

Lithium salts

Quality Level

Assay

>98% (HPLC)

packaging

pkg of 4 × 200 μL (4 x 20 μmol; 100 mM, each)

manufacturer/tradename

Roche

technique(s)

DNA sequencing: suitable

storage temp.

−20°C

General description

Ribonucleoside Triphosphate Set consists of four separate vials of ATP, CTP, GTP, and UTP. Each vial contains a 100 mM clear, colorless solution of the lithium salt (pH 7).

Application

The Ribonucleoside Triphosphate Set is suitable for use in in vitro-transcription reactions and RNA polymerase-directed DNA sequencing.

Quality

Typical analysis:
≥98% ribonucleoside triphosphates (HPLC), ≤1.5% ribonucleoside diphosphates, ≤0.5% ribonucleoside monophosphates.
Tested for the absence of RNases according to the current Quality Control procedures.

Unit Definition

1 mM ATP = 15.3 A260 units, 1 mM CTP = 9.6 A272 units, 1 mM GTP = 13.7 A252 units, 1 mM UTP = 9.9 A260 units.

Other Notes

For life science research only. Not for use in diagnostic procedures. For your convenience, ribonucleotides are also available as single NTPs.
Ribonucleotides are also included in some kits (NTPs, buffer, and enzyme in separate vials).

Kit Components Only

Product No.
Description

  • ATP, lithium salt 100 mM

  • CTP, lithium salt 100 mM

  • GTP, lithium salt 100 mM

  • UTP, lithium salt 100 mM

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiang Ning et al.
Nature communications, 12(1), 5832-5832 (2021-10-07)
Maize ear size and kernel number differ among lines, however, little is known about the molecular basis of ear length and its impact on kernel number. Here, we characterize a quantitative trait locus, qEL7, to identify a maize gene controlling
Philip B Brewer et al.
Nature protocols, 1(3), 1462-1467 (2007-04-05)
Plant biology is currently confronted with an overflow of expression profile data provided by high-throughput microarray transcription analyses. However, the tissue and cellular resolution of these techniques is limited. Thus, it is still necessary to examine the expression pattern of
Adam D Langenbacher et al.
EvoDevo, 7, 9-9 (2016-04-14)
Germ cells are specified during early development and are responsible for generating gametes in the adult. After germ cells are specified, they typically migrate to a particular niche in the organism where they reside for the remainder of its lifetime.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service