Skip to Content
Merck
All Photos(5)

Key Documents

U6625

Sigma-Aldrich

Uridine 5′-triphosphate trisodium salt hydrate

from yeast, Type III, ≥96%

Synonym(s):

UTP Na3

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$45.30
100 MG
$151.00
500 MG
$491.00
1 G
$839.00

$45.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$45.30
100 MG
$151.00
500 MG
$491.00
1 G
$839.00

About This Item

Empirical Formula (Hill Notation):
C9H12N2Na3O15P3 · xH2O
CAS Number:
Molecular Weight:
550.09 (anhydrous basis)
Beilstein:
3585234
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$45.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

yeast

type

Type III

Assay

≥96%

solubility

H2O: soluble 50 mg/mL, clear to slightly hazy, colorless

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])(=O)OP([O-])(=O)OP(O)([O-])=O)N2C=CC(=O)NC2=O

InChI

1S/C9H15N2O15P3.3Na/c12-5-1-2-11(9(15)10-5)8-7(14)6(13)4(24-8)3-23-28(19,20)26-29(21,22)25-27(16,17)18;;;/h1-2,4,6-8,13-14H,3H2,(H,19,20)(H,21,22)(H,10,12,15)(H2,16,17,18);;;/q;3*+1/p-3/t4-,6-,7-,8-;;;/m1.../s1

InChI key

MMJGIWFJVDOPJF-LLWADOMFSA-K

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

UTP has been used to study its vasodilatory effects on the skeletal muscle cells of diabetic patients[1]. UTP has also been used as a purinergic agonist to study ion transport functions of P2Y2 receptors on rat basolateral membranes[2].

Biochem/physiol Actions

P2Y receptor agonist.

Preparation Note

Uridine 5′-triphosphate trisodium salt (UTP Na3) hydrate dissolves in water at 50 mg/ml to yield a clear to slightly hazy, colorless solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satyanarayana R Pondugula et al.
BMC research notes, 3, 100-100 (2010-04-20)
The ionic composition of the luminal fluid in the vestibular labyrinth is maintained within tight limits by the many types of epithelial cells bounding the lumen. Regulatory mechanisms include systemic, paracrine and autocrine hormones along with their associated intracellular signal
Xiping Chen et al.
FEBS letters, 595(1), 110-122 (2020-10-25)
O-GlcNAcylation is a post-translational modification catalysed by O-GlcNAc transferase (OGT). Missense mutations in OGT have been associated with developmental disorders, OGT-linked congenital disorder of glycosylation (OGT-CDG), which are characterized by intellectual disability. OGT relies on the hexosamine biosynthetic pathway (HBP)
Alexander Puzenko et al.
The Journal of chemical physics, 137(19), 194502-194502 (2012-11-28)
In this, the third part of our series on the dielectric spectrum symmetrical broadening of water, we consider the nucleotide aqueous solutions. Where in Parts I [E. Levy et al., J. Chem. Phys. 136, 114502 (2012)] and II [E. Levy
Yinlong Yang et al.
Cell reports, 32(12), 108161-108161 (2020-09-24)
Sensing stressful conditions and adjusting the cellular metabolism to adapt to the environment are essential activities for bacteria to survive in variable situations. Here, we describe a stress-related protein, YdiU, and characterize YdiU as an enzyme that catalyzes the covalent
Lilan You et al.
Cell, 176(1-2), 239-253 (2018-12-07)
Csm, a type III-A CRISPR-Cas interference complex, is a CRISPR RNA (crRNA)-guided RNase that also possesses target RNA-dependent DNase and cyclic oligoadenylate (cOA) synthetase activities. However, the structural features allowing target RNA-binding-dependent activation of DNA cleavage and cOA generation remain

Protocols

HILIC separation is an alternative that permits sensitive MS detection and without the use of ion-pair reagents.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service