Skip to Content
Merck
All Photos(2)

Key Documents

T1699

Sigma-Aldrich

Trizma® base solution

1.5 M, liquid

Synonym(s):

2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol, Tris base solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$59.10

$59.10


Estimated to ship onApril 08, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$59.10

About This Item

CAS Number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.25

$59.10


Estimated to ship onApril 08, 2025


Request a Bulk Order

Product Name

Trizma® base solution, 1.5 M

form

liquid

Quality Level

concentration

1.5 M

pH

10.7-11.1

application(s)

diagnostic assay manufacturing

storage temp.

2-8°C

SMILES string

NC(CO)(CO)CO

InChI

1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2

InChI key

LENZDBCJOHFCAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For use in the enzymatic determination of pyruvate using lactic dehydrogenase.

Caution

Contains 0.05% sodium azide as a preservative.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Uma Kavita et al.
Journal of immunological methods, 448, 91-104 (2017-06-20)
We developed a homogeneous bridging anti-drug antibody (ADA) assay on an electro chemiluminescent immunoassay (ECLIA) platform to support the immunogenicity evaluation of a dimeric domain antibody (dAb) therapeutic in clinical studies. During method development we evaluated the impact of different
Satoshi Hokari et al.
Molecular oncology, 14(2), 277-293 (2019-11-30)
Thyroid transcription factor-1 (TTF-1, encoded by the NKX2-1 gene) is highly expressed in small-cell lung carcinoma (SCLC) and lung adenocarcinoma (LADC), but how its functional roles differ between SCLC and LADC remains to be elucidated. Here, we compared the genome-wide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service