Skip to Content
Merck
All Photos(1)

Documents

1.00049

Supelco

Calcium Test, photometric

photometric, 0.20-4.00 mg/L (Ca), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Calcium Test, photometric, 0.20-4.00 mg/L (Ca), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 100 tests

specific analyte(s)

calcium

measuring range

0.20-4.00 mg/L (Ca)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Phthalein derivate)

storage temp.

15-25°C

General description

Method: photometric 0.20 - 4.00 mg/l Ca Spectroquant®

This Spectroquant® Calcium Reagent Test allows the accurate quantification of the calcium content in aqueous samples.

Method applied: In alkaline solution calcium ions react with a phthalein derivative to form a violet dye which is determined photometrically. The 8-hydroxyquinoline contained in reagent Ca-1 prevents interferences by magnesium and iron.

The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.

All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Met. Corr. 1 - Repr. 1B

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Protocols

Using the Spectroquant® photometric system, we outline a photometric analysis of calcium levels in beer.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service