Skip to Content
Merck
All Photos(8)

Documents

173018

Supelco

Prove 600 Spectrophotometer

suitable for UV/Vis spectroscopy, Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
SE.11

product name

Prove 600, suitable for UV/Vis spectroscopy, Spectroquant®

product line

Spectroquant®

Quality Level

technique(s)

UV/Vis spectroscopy: suitable
photometry: suitable

compatibility

for use with Spectroquant® Prove 600

shipped in

ambient

storage temp.

no temp limit

General description

Equipped with high-end UV/Vis optics, and the capability to hold cells of up to 100 mm, the Spectroquant Prove 600 packs great power into a compact size. The Spectroquant Prove 600 delivers excellent resolution and sensitivity for use with test kits, complex kinetics or spectral measurements.

The Spectroquant Prove family is our series of cutting edge spectrophotometers for easy analysis of water, food and beverage, and environmental samples. All Spectroquant Prove instruments are pre-programmed for over 200 Test Kits and automatic bar code recognition ensures the correct method parameters are applied and all essential information such as lot number, expiry date, and automatic calibration updates are recorded, making the use of analytical Test Kits as easy as never before.
UV/VIS spectrophotometer 1,8 nm spectral bandwidth Spectroquant®

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Articles

We are a global leader in the life science industry and have produced test kits to measure numerous analytes. Water Online spoke with us about advances in measuring chemical oxygen demand.

Learn about the UV-spectrometric determination of the K270 nm value of olive oil, assessing the UV absorbance at 270 nm to ensure quality and compliance with regulatory standards.

Learn about the UV-spectrometric determination of the K268 nm value of olive oil, assessing the UV absorbance at 268 nm, to ensure quality and compliance with regulatory standards.

Spectroquant® photometric test accurately quantifies low concentrations of PO4-P in natural waters.

See All

Protocols

Ammonium is the naturally preferred source of nitrogen for plant growth and there is an increasing global demand for organic food products.

Using the Spectroquant® photometric system, we outline a photometric analysis of calcium levels in beer.

Related Content

Learn about the sample preparation and photometric determination of non-ionic surfactants in Extran® rinse solutions (MA 01, MA 02, MA 05, AP 22) using Spectroquant® test kits and spectrophotometers.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service