Skip to Content
Merck
All Photos(8)

Documents

173016

Supelco

Prove 100 Spectrophotometer

Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
SE.11

product name

Prove 100, Spectroquant®

product line

Spectroquant®

Quality Level

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Prove 100

shipped in

ambient

storage temp.

no temp limit

General description

The Spectroquant Prove 100 is the best choice for those who primarily use our broad range of Spectroquant Test Kits, or only perform Vis measurements.

The Spectroquant Prove 100 provides high quality and great value for money for your daily analyses.

The Spectroquant Prove family is our series of cutting edge spectrophotometers for easy analysis of water, food and beverage, and environmental samples. All Spectroquant Prove instruments are pre-programmed for over 200 Test Kits and automatic bar code recognition ensures the correct method parameters are applied and all essential information such as lot number, expiry date, and automatic calibration updates are recorded, making the use of analytical Test Kits as easy as never before.
VIS Spectrophotometer 4 nm spectral bandwidth Spectroquant®

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

We are a global leader in the life science industry and have produced test kits to measure numerous analytes. Water Online spoke with us about advances in measuring chemical oxygen demand.

Urea in water and wastewater, enzymatic determination - Photometric determination using the indophenol blue method after cleavage of the urea

Total Nitrogen Analysis kit offers a fast, accurate, and cost-effective method for water and wastewater testing.

A step-by-step procedure for the photometric determination of Nitrate in effluents using Spectroquant® test kits and photometer for a precise and quantitative analysis.

Protocols

Ammonium is the naturally preferred source of nitrogen for plant growth and there is an increasing global demand for organic food products.

Using the Spectroquant® photometric system, we outline a photometric analysis of calcium levels in beer.

Method: Photometric determination with 2,2'-dinitro-5,5'-dithiodibenzoic acid (Ellman's reagent)

Photometric determination with PAR subsequent to acid mineralisation; DIN decomposition

See All

Related Content

Learn about the sample preparation and photometric determination of non-ionic surfactants in Extran® rinse solutions (MA 01, MA 02, MA 05, AP 22) using Spectroquant® test kits and spectrophotometers.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service