Skip to Content
Merck
All Photos(1)

Documents

1.01758

Supelco

Acid Capacity Cell Test to pH 4.3 (total alkalinity)

photometric, 0.40-8.00 mmol/L (CaCO3), 20-400 mg/L (CaCO3), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product line

Spectroquant®

Quality Level

usage

sufficient for 120 tests

analyte functional class(es)

acid capacity

measuring range

0.40-8.00 mmol/L (CaCO3)
20-400 mg/L (CaCO3)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Indicator)

storage temp.

15-25°C

General description

Acid capacity or carbonate hardness/total alkalinity refers to the buffering capacity of a solution against acidification. It depends on the concentration of hydrocarbonates and carbonates. These react in the test with an indicator, forming a blue color that is determined photometrically.The Spectroquant® Cell Tests come with prefilled 16 mm round cells as well as all the required reagents to perform the analysis according to the procedure described in the accompanying instruction leaflet.All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service