Skip to Content
Merck
All Photos(1)

Key Documents

860517P

Avanti

C17 Ceramide (d18:1/17:0)

Avanti Research - A Croda Brand

Synonym(s):

N-heptadecanoyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$130.00
10 MG
$243.00
25 MG
$507.00

$130.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$130.00
10 MG
$243.00
25 MG
$507.00

About This Item

Empirical Formula (Hill Notation):
C35H69NO3
CAS Number:
Molecular Weight:
551.93
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$130.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

form

powder

packaging

pkg of 1 × 10 mg (860517P-10mg)
pkg of 1 × 25 mg (860517P-25mg)
pkg of 1 × 5 mg (860517P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCCCCCCCCCCC)=O)[C@]([H])(O)/C=C/CCCCCCCCCCCCC

InChI

1S/C35H69NO3/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-35(39)36-33(32-37)34(38)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h28,30,33-34,37-38H,3-27,29,31-32H2,1-2H3,(H,36,39)/b30-28+/t33-,34+/m0/s1

InChI key

ICWGMOFDULMCFL-QKSCFGQVSA-N

General description

Ceramide synthesis is mediated either by the salvage pathway by the acylation of sphingosine or sphingolipids hydrolysis or by de novo pathway via dihydroceramide formation.[1] Structurally, ceramides have a sphingoid base, a long-chain amino alcohol linked to fatty acid by an amide bond.[2]

Application

C17 Ceramide (d18:1/17:0) has been used:
  • as an internal standard for myocardial ceramide quantification using chromatography with negative ion electrospray ionization coupled to tandem mass spectrometry[3]
  • as an internal standard in tandem mass spectrometry (MS/MS)[4]
  • as a substrate for sphingomyelin synthase 2 enzyme[5]

Biochem/physiol Actions

Ceramides play a key role in regulating various cellular processes like senescence and apoptosis.[1] They are directly involved in the proliferation and differentiation of skin cells and regulate skin barrier functionality.[2] Ceramide based analogs are potential anti-tumor agents and are regarded as tumor suppressor lipid.[6]

Packaging

5 mL Amber Glass Screw Cap Vial (860517P-10mg)
5 mL Amber Glass Screw Cap Vial (860517P-25mg)
5 mL Amber Glass Screw Cap Vial (860517P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ye-Ryung Kim et al.
Experimental & molecular medicine, 51(11), 129-129 (2019-11-05)
The endoplasmic reticulum (ER) is not only important for protein synthesis and folding but is also crucial for lipid synthesis and metabolism. In the current study, we demonstrate an important role of ceramide synthases (CerS) in ER stress and NAFLD
Takuya Takeichi et al.
The Journal of clinical investigation, 130(2), 890-903 (2019-11-02)
The corneocyte lipid envelope, composed of covalently bound ceramides and fatty acids, is important to the integrity of the permeability barrier in the stratum corneum, and its absence is a prime structural defect in various skin diseases associated with defective
André F Nascimento et al.
Life sciences, 88(23-24), 1031-1038 (2011-04-05)
Leptin resistance has been associated with cardiac lipotoxicity; however, whether leptin resistance is a risk factor associated with cardiac lipotoxicity at different time points in diet-induced obesity is unclear. The objective of this study was to evaluate this relationship. Male
Long-term high-fat diet-induced obesity decreases the cardiac leptin receptor without apparent lipotoxicity
Nascimento AF, et al.
Life Sciences, 88(23-24), 1031-1038 (2011)
Prosaposin deficiency and saposin B deficiency (activator-deficient metachromatic leukodystrophy): report on two patients detected by analysis of urinary sphingolipids and carrying novel PSAP gene mutations
Kuchar L, et al.
American Journal of Medical Genetics. Part A, 149(4), 613-621 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service