Skip to Content
Merck
All Photos(1)

Key Documents

PHR1512

Supelco

Ergosterol

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Ergosterol, 3β-Hydroxy-5,7,22-ergostatriene, 5,7,22-Ergostatrien-3β-ol, Provitamin D2

Sign Into View Organizational & Contract Pricing

Select a Size

200 MG
$154.00

$154.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 MG
$154.00

About This Item

Empirical Formula (Hill Notation):
C28H44O
CAS Number:
Molecular Weight:
396.65
Beilstein:
2338604
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$154.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. E1100000
traceable to USP 1241007

API family

ergosterol

form

solid

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

156-158 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]1(CC[C@@]2([H])C3=CC=C4C[C@@H](O)CC[C@]4(C)[C@@]3([H])CC[C@]12C)[C@H](C)\C=C\[C@H](C)C(C)C

InChI

1S/C28H44O/c1-18(2)19(3)7-8-20(4)24-11-12-25-23-10-9-21-17-22(29)13-15-27(21,5)26(23)14-16-28(24,25)6/h7-10,18-20,22,24-26,29H,11-17H2,1-6H3/b8-7+/t19-,20+,22-,24+,25-,26-,27-,28+/m0/s1

InChI key

DNVPQKQSNYMLRS-APGDWVJJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ergosterol is a biological precursor of Vitamin D2 found in cell membranes of fungi and some protists such as trypanosomes. It is a major sterol found in fungal cell membranes.[1]
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA3319 in the slot below. This is an example certificate only and may not be the lot that you receive.

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volha Shapaval et al.
Biotechnology for biofuels, 12, 140-140 (2019-06-11)
Oleaginous yeasts are considered as a potential lipid source for food, feed and biofuel production. In order to make the yeast-based lipid production environmentally and economically sustainable, there is a need for screening studies in order to find the best
Analysis of ergosterol in single kernel and ground grain by gas chromatography- mass spectrometry
Dong Y, et al.
Journal of Agricultural and Food Chemistry, 54(12), 4121-4125 (2006)
Leo T Mpofu et al.
Planta, 240(2), 239-250 (2014-05-13)
A lack of understanding of host-by-pathogen relations can hinder the success of breeding for resistance to a major disease. Fungal strain pathogenicity has to be understood from the virulence it can cause on susceptible genotypes and host resistance indicates which
Benoît Pilmis et al.
The Journal of antimicrobial chemotherapy, 70(1), 14-22 (2014-09-11)
Antifungal prescription remains a challenge in pregnant women because of uncertainties regarding fetal toxicity and altered maternal pharmacokinetic parameters that may affect efficacy or increase maternal and fetal toxicity. We present updated data reviewing the available knowledge and current recommendations
Wei-Jie Wu et al.
PloS one, 9(4), e95359-e95359 (2014-04-17)
Response surface methodology (RSM) was used to determine the optimum vitamin D2 synthesis conditions in oyster mushrooms (Pleurotus ostreatus). Ultraviolet B (UV-B) was selected as the most efficient irradiation source for the preliminary experiment, in addition to the levels of

Questions

  1. If I make solutions from the product, at what condition should I storage it? Should it also be storage at 4-8°C as it's powdered form?

    1 answer
    1. The stability of this product in solution has not been evaluated. However, product 47130-U is a 10 mg/mL solution of Ergosterol in chloroform. This product is stored at 2-8°C and is assigned a shelf life of 1 year.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service