Skip to Content
Merck
All Photos(2)

Key Documents

860646P

Avanti

C18 Ceramide (d17:1/18:0)

Avanti Research - A Croda Brand 860646P, powder

Synonym(s):

N-stearoyl-D-erythro-sphingosine (C17 base)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$994.00

$994.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$994.00

About This Item

Empirical Formula (Hill Notation):
C35H69NO3
CAS Number:
Molecular Weight:
551.93
UNSPSC Code:
12352211
NACRES:
NA.25

$994.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 10 mg (860646P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860646P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCCCCCCCCCCCC)=O)[C@]([H])(O)/C=C/CCCCCCCCCCCC

InChI key

KGEXFGDEDXQZLQ-QKSCFGQVSA-N

General description

Ceramide is a bioactive sphingolipid. C18 Ceramide (d17:1/18:0) is synthesized by the N-acylation of sphingosine.[1] This reaction is catalyzed by ceramide synthase 1 (CerS1).[2] C18 Ceramide is highly expressed in brain tissues.[1]

Application

C18 Ceramide (d17:1/18:0) has been used as an internal standard in mass spectrometer and ultra high performance Liquid Chromatography for quantitative measurement of ceramides in muscle.[3]

Biochem/physiol Actions

C18 Ceramide has an ability to induce cell death.[4] Defects in the generation or accumulation of C18 Ceramide has been associated with the development of human head and neck squamous cell carcinomas (HNSCC).[2]

Packaging

5 mL Amber Glass Screw Cap Vial (860646P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The structure of D-erythro-C18 ceramide at the air-water interface
Vaknin D and Kelley MS
Biophysical Journal, 79(5), 2616-2623 (2000)
Clinical relevance of ceramide metabolism in the pathogenesis of human head and neck squamous cell carcinoma (HNSCC): attenuation of C18-ceramide in HNSCC tumors correlates with lymphovascular invasion and nodal metastasis
Karahatay S, et al.
Cancer Letters, 256(1), 101-111 (2007)
Long-term follow-up of muscle lipid accumulation, mitochondrial activity and oxidative stress and their relationship with impaired glucose homeostasis in high fat high fructose diet-fed rats
Wang Y, et al.
The Journal of Nutritional Biochemistry, 64(5), 182-197 (2019)
Koichi Ishida et al.
The Journal of investigative dermatology (2020-02-06)
Little is known about the pathophysiological linkages between altered ceramide profiles in the stratum corneum (SC) of patients with atopic dermatitis and their impaired skin barrier and water-holding functions. We studied those characteristics following topical treatment with a designed synthetic
Diverse functions of ceramide in cancer cell death and proliferation
Advances in Cancer Research, 117, 37-58 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service