Skip to Content
Merck
All Photos(2)

Key Documents

830856P

Avanti

17:0 PA

1,2-diheptadecanoyl-sn-glycero-3-phosphate (sodium salt), powder

Synonym(s):

PA(17:0/17:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$253.00

$253.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$253.00

About This Item

Empirical Formula (Hill Notation):
C37H72O8PNa
CAS Number:
Molecular Weight:
698.93
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$253.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (830856P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 830856P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCCCCCCCCCCC)COC(=O)CCCCCCCCCCCCCCCC)O

InChI

1S/C37H73O8P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-36(38)43-33-35(34-44-46(40,41)42)45-37(39)32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h35H,3-34H2,1-2H3,(H2,40,41,42);/q;+1/p-1/t35-;/m1./s1

InChI key

IAELRFKIVVBNTO-RUQJKXHKSA-M

General description

Phosphatidic acid (PA), a phospholipid is a minor component of the phospholipid pool. It is made up of two fatty acids and a phosphate group and are covalently attached with the help of ester linkage to a glycerol molecule. PA is an important constituent of cell membranes.[1]

Application

17:0 PA has been used as an internal standard for BXD liver samples/lipidomics analysis[2] and as an internal standard for BXD plasma sample analysis.[3] It may be used as a component in internal standard mix to spike the brain tissue homogenate.[4]

Biochem/physiol Actions

Phosphatidic acid (PA) serves as a lipid messenger. It can enhance the production of muscle protein. PA can also serve as a signaling lipid. It plays a key role in mammalian target of rapamycin (mTOR) signaling.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (830856P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sophie Ayciriex et al.
Frontiers in molecular neuroscience, 10, 211-211 (2017-07-27)
Impairment in cholesterol metabolism is associated with many neurodegenerative disorders including Alzheimer's disease (AD). However, the lipid alterations underlying neurodegeneration and the connection between altered cholesterol levels and AD remains not fully understood. We recently showed that cholesterol accumulation in
Yoshibumi Shimizu et al.
Prostaglandins & other lipid mediators, 150, 106450-106450 (2020-04-17)
Cyclic phosphatidic acid (cPA) is a lysophospholipid mediator that suppresses cancer metastasis and osteoarthritis. It also has neuroprotective roles in diseases such as multiple sclerosis and delayed neuronal death following transient ischemia. In order to take advantage of the properties
Emily L Gill et al.
Journal of proteome research, 19(1), 424-431 (2019-11-13)
Parkinson's disease (PD) is characterized by the loss of dopaminergic neurons in the substantia nigra pars compacta of the brain, as well as the degeneration of motor and nonmotor circuitries. The cause of neuronal death is currently unknown, although chronic
Pooja Jha et al.
Cell systems, 6(6), 722-733 (2018-06-18)
The genetics of individual lipid species and their relevance in disease is largely unresolved. We profiled a subset of storage, signaling, membrane, and mitochondrial liver lipids across 385 mice from 47 strains of the BXD mouse population fed chow or
Guillermo Escalante et al.
Journal of the International Society of Sports Nutrition, 13, 24-24 (2016-06-09)
Phosphatidic acid (PA) is a lipid messenger that has been shown to increase muscle protein synthesis via signaling stimulation of the mammalian target of rapamycin (mTOR). MaxxTOR® (MT) is a supplement that contains PA as the main active ingredient but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service