Skip to Content
Merck
All Photos(1)

Key Documents

860473P

Avanti

1-desoxymethylsphinganine

Avanti Research - A Croda Brand 860473P, powder

Synonym(s):

1-desoxymethylsphinganine (m17:0)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$405.00

$405.00


Estimated to ship onApril 10, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$405.00

About This Item

Empirical Formula (Hill Notation):
C17H37NO
CAS Number:
Molecular Weight:
271.48
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$405.00


Estimated to ship onApril 10, 2025


Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (860473P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860473P

lipid type

bioactive lipids
sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

NC[C@]([H])(O)CCCCCCCCCCCCCCC

InChI key

UGVBFHUWZNNKIK-QGZVFWFLSA-N

General description

1-desoxymethylsphinganine is synthesized by the condensation of alanine with palmitoyl-CoA.[1]

Application

1-desoxymethylsphinganine is suitable for use for testing its cytotoxic effects on primary neuron-glia cultures.[2] It is also suitable for use as a neurotoxic agent in human CD8+ T cells.[3] In hereditary sensory and autonomic neuropathy type I, mutations in serine palmitoyltransferase long chain base subunit 1 resulting in altered substrate specificity and accumulation of 1.[4]

Biochem/physiol Actions

1-desoxymethylsphinganine is neurotoxic towards CD8+ T cells and inhibits cytokine production.[3] However, it is not cytotoxic to dopaminergic neurons.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (860473P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mutations in the SPTLC2 subunit of serine palmitoyltransferase cause hereditary sensory and autonomic neuropathy type I
Rotthier A, et al.
American Journal of Human Genetics, 87(4), 513-522 (2010)
Loss of Neurological Disease HSAN-I-Associated Gene SPTLC2 Impairs CD8+ T Cell Responses to Infection by Inhibiting T Cell Metabolic Fitness
Wu J, et al.
Immunity, 50(5), 1218-1231 (2019)
Biophysical properties of novel 1-deoxy-(dihydro) ceramides occurring in mammalian cells
Jimenez-Rojo N, et al.
Biophysical Journal, 107(12), 2850-2859 (2014)
Ceramide sphingolipid signaling mediates Tumor Necrosis Factor (TNF)-dependent toxicity via caspase signaling in dopaminergic neurons
Martinez TN, et al.
Mol. Neurodegener., 7(1), 45-45 (2012)
Nicholas C Zitomer et al.
The Journal of biological chemistry, 284(8), 4786-4795 (2008-12-20)
Fumonisin B(1) (FB(1)) is a mycotoxin that inhibits ceramide synthases (CerS) and causes kidney and liver toxicity and other disease. Inhibition of CerS by FB(1) increases sphinganine (Sa), Sa 1-phosphate, and a previously unidentified metabolite. Analysis of the latter by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service