Skip to Content
Merck
All Photos(2)

Key Documents

855475P

Avanti

12:0 Lyso PC

1-lauroyl-2-hydroxy-sn-glycero-3-phosphocholine, powder

Synonym(s):

1-dodecanoyl-sn-glycero-3-phosphocholine; PC(12:0/0:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$216.00
200 MG
$405.00

$216.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$216.00
200 MG
$405.00

About This Item

Empirical Formula (Hill Notation):
C20H42NO7P
CAS Number:
Molecular Weight:
439.52
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$216.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

powder

packaging

pkg of 1 × 200 mg (855475P-200mg)
pkg of 1 × 25 mg (855475P-25mg)
pkg of 1 × 500 mg (855475P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855475P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COC(CCCCCCCCCCC)=O

InChI

1S/C20H42NO7P/c1-5-6-7-8-9-10-11-12-13-14-20(23)26-17-19(22)18-28-29(24,25)27-16-15-21(2,3)4/h19,22H,5-18H2,1-4H3/t19-/m1/s1

InChI key

BWKILASWCLJPBO-LJQANCHMSA-N

General description

Lyso-phosphatidylcholine (LPC) is a bioactive proinflammatory lipid secreted on pathological response.[1] It has a choline group at its polar head.[2] Lysophosphatidylcholine (Lyso-PC) is a component of blood plasma, vascular tissue and lipoproteins.[3]

Application

12:0 Lyso PC has been used to study its neuritogenic activity[4] and the applications of exogenous lipids.[5]

Biochem/physiol Actions

Lysophosphatidylcholine (LPC) alters various functions in different cell-types, like endothelial cells, smooth muscle cells, monocytes, macrophages and T-cells. It is associated with atherosclerosis and inflammatory diseases. LPC activates many second messengers including, protein kinase C, extracellular-signal-regulated kinases and protein tyrosine kinases.[1]

Packaging

20 mL Clear Glass Screw Cap Vial (855475P-500mg)
5 mL Amber Glass Screw Cap Vial (855475P-200mg)
5 mL Amber Glass Screw Cap Vial (855475P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inner/outer nuclear membrane fusion in nuclear pore assembly: biochemical demonstration and molecular analysis
Fichtman B, et al.
Molecular Biology of the Cell, 21(23) (2010)
Secretory phospholipases A2 induce neurite outgrowth in PC12 cells through lysophosphatidylcholine generation and activation of G2A receptor
Ikeno Y, et al.
Test, 280(30) (2005)
Boris Fichtman et al.
Molecular biology of the cell, 21(23), 4197-4211 (2010-10-12)
Nuclear pore complexes (NPCs) are large proteinaceous channels embedded in double nuclear membranes, which carry out nucleocytoplasmic exchange. The mechanism of nuclear pore assembly involves a unique challenge, as it requires creation of a long-lived membrane-lined channel connecting the inner
Shi-Yuan Wen et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 69, 152690-152690 (2018-11-06)
Rhodioloside is a glucoside of tyrosol isolated from Rhodiola rosea. However, its regulating effect on hepatic dyslipidemia of atherogenic mice has rarely been studied. The specific aims of current study included to clarify lipidomic perturbation in liver tissues of apolipoprotein
Lysophosphatidylcholine stimulates the release of arachidonic acid in human endothelial cells
Wong JT, et al.
Test, 273(12), 6830-6836 (1998)

Articles

The critical micelle concentration (CMC) can be approximately defined as the lipid monomer concentration at which appreciable amounts (>5% of total) of micellar aggregates first begin to appear in the equilibrium: nM1<=>Mn

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service