Skip to Content
Merck
All Photos(1)

Key Documents

860477P

Avanti

1-desoxymethylsphingosine

1-desoxymethylsphingosine (m17:1), powder

Synonym(s):

1-Deoxymethylsphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$405.00

$405.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$405.00

About This Item

Empirical Formula (Hill Notation):
C17H35NO
CAS Number:
Molecular Weight:
269.47
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$405.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (860477P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860477P

lipid type

sphingolipids
bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC/C=C/[C@](O)([H])CN

InChI

1S/C17H35NO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-17(19)16-18/h14-15,17,19H,2-13,16,18H2,1H3/b15-14+/t17-/m1/s1

InChI key

NTZGVZKEAIALQE-IAOULYHDSA-N

General description

1-desoxymethylsphingosine is synthesized from glycine in the presence of the enzyme serine palmitoyltransferase (SPT).[1]

Application

1-desoxymethylsphingosine may be used for the complex preparation with bovine serum albumin for cytotoxicity testing in MN9D dopaminergic neuroblastoma cell line.[2]

Biochem/physiol Actions

1-desoxymethylsphingosine is less neurotoxic compared to 1-desoxysphingosine in dorsal root ganglion neuronscultures[3] Mutations in the palmitoyltransferase (SPT) in hereditary sensory neuropathy type 1 (HSAN-1), due to altered substrate specificity 1-desoxymethylsphinganine and 1-deoxy-sphinganine which are further converted to -deoxy(methyl)-ceramide and 1-deoxy(methyl)-sphingosine (1-deoxy(methyl)-SO). However, they lack the hydroxyl group essential for glycosphingolipid generation, resulting in accumulation of intermediate products.[4]

Packaging

5 mL Amber Glass Screw Cap Vial (860477P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1-Deoxysphingolipids encountered exogenously and made de novo: dangerous mysteries inside an enigma
Duan, Jingjing and Merrill, Alfred H
The Journal of Biological Chemistry, 290(25), 15380-15389 (2015)
Annelies Rotthier et al.
Human mutation, 32(6), E2211-E2225 (2011-05-28)
Hereditary sensory and autonomic neuropathy type I (HSAN-I) is an axonal peripheral neuropathy leading to progressive distal sensory loss and severe ulcerations. Mutations in SPTLC1 and SPTLC2, encoding the two subunits of serine palmitoyltransferase (SPT), the enzyme catalyzing the first
Ceramide sphingolipid signaling mediates Tumor Necrosis Factor (TNF)-dependent toxicity via caspase signaling in dopaminergic neurons
Martinez TN, et al.
Mol. Neurodegener., 7(1), 45-45 (2012)
Hereditary sensory neuropathy type 1 is caused by the accumulation of two neurotoxic sphingolipids
Penno A, et al.
The Journal of Biological Chemistry, 285(15), 11178-11187 (2010)
Anke Penno et al.
The Journal of biological chemistry, 285(15), 11178-11187 (2010-01-26)
HSAN1 is an inherited neuropathy found to be associated with several missense mutations in the SPTLC1 subunit of serine palmitoyltransferase (SPT). SPT catalyzes the condensation of serine and palmitoyl-CoA, the initial step in the de novo synthesis of sphingolipids. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service