Skip to Content
Merck
All Photos(1)

Key Documents

171204C

Avanti

Heart Extract Polar

Avanti Research - A Croda Brand

Synonym(s):

Heart Polar Lipid Extract (Bovine)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$284.00

$284.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$284.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$284.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

form

chloroform solution

packaging

pkg of 1 × 4 mL (171204C-100mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

25 mg/mL (171204C-100mg)

lipid type

lipid extracts

shipped in

dry ice

storage temp.

−20°C

General description

Heart Extract Polar is a natural lipid. Polar Lipid Extract is derived from the total lipid extract by precipitation with acetone followed by extraction of the acetone insoluble material with diethyl ether.

Application

Heart Extract Polar is suitable for use
  • as a component of lipid mixture for mimicking mammalian cellular membrane and in synchrotron small-angle X-ray diffraction measurements[1]
  • for spiking mobilograms for phosphaltidyl choline 18:1/18:1 in ion-mobility spectrometry- time of flight measurements(IMS-TOF)[2]
  • in the preparation of large unilamellar vesicles[3]

Packaging

5 mL Clear Glass Sealed Ampule (171204C-100mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

High resolution ion mobility-mass spectrometry for separation and identification of isomeric lipids
Groessl M, et al.
Analyst, 140(20), 6904-6911 (2015)
Inorganic mercury and cadmium induce rigidity in eukaryotic lipid extracts while mercury also ruptures red blood cells
Kerek E, et al.
Biochimica et Biophysica Acta - Biomembranes, 1860(3), 710-717 (2018)
Natural lipid extracts and biomembrane-mimicking lipid compositions are disposed to form nonlamellar phases, and they release DNA from lipoplexes most efficiently
Koynova R and MacDonald RC
Biochimica et Biophysica Acta - Biomembranes, 1768(10), 2373-2382 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service