Skip to Content
Merck
All Photos(1)

Key Documents

171201P

Avanti

Heart Extract Total

Avanti Research - A Croda Brand

Synonym(s):

Heart Total Extract (Bovine)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$158.00
1 G
$714.00

$158.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$158.00
1 G
$714.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$158.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 g (171201P-1g)
pkg of 1 × 100 mg (171201P-100mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

lipid extracts

shipped in

dry ice

storage temp.

−20°C

General description

Total Lipid Extract is a chloroform:methanol extract of the respective tissue (Brain, Soybean, Heart, etc.). This extract is partitioned against deionized water, and the chloroform phase is concentrated.

Packaging

5 mL Clear Glass Sealed Ampule (171201P-100mg)
60 mL Amber Wide Mouth Screw Cap Glass Bottle (171201P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hari Kiran Kotapati et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1145, 122099-122099 (2020-04-20)
Three normal phase HPLC methods were produced to separate lipid classes on a PVA-Sil stationary phase including: 9 polar lipids (method 1); 13 combined polar and neutral lipids (method 2); and a combined method that further separates the neutral lipids
Giuseppe Paglia et al.
Nature protocols, 12(4), 797-813 (2017-03-17)
Metabolomics and lipidomics aim to profile the wide range of metabolites and lipids that are present in biological samples. Recently, ion mobility spectrometry (IMS) has been used to support metabolomics and lipidomics applications to facilitate the separation and the identification

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service