Skip to Content
Merck
All Photos(1)

Key Documents

141101P

Avanti

Brain Extract Polar

Avanti Research - A Croda Brand

Synonym(s):

Brain Polar Lipid Extract (Porcine)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$284.00
500 MG
$879.00

$284.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$284.00
500 MG
$879.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$284.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

form

powder

packaging

pkg of 1 × 100 mg (141101P-100mg)
pkg of 5 × 100 mg (141101P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

lipid extracts

shipped in

dry ice

storage temp.

−20°C

General description

Brain Extract Polar comprises phosphatidylethanolamine as a major lipid component. It also contains phosphatidylserine and phosphatidylcholine. The minor components include the phosphatidic acid and phosphatidylinositol.
Brain Polar Lipid Extract is derived from the total lipid extract by precipitation with acetone followed by extraction of the acetone insoluble material with diethyl ether.

Application

Brain Extract Polar has been used in parallel artificial membrane permeability assay (PAMPA) for coating membrane to evaluate blood brain-barrier permeability,[1][2] and for drug permeability studies.[3]

Biochem/physiol Actions

Lipid extracts from the mammalian tissues like brain, kidney and liver are used in generating model membranes for mimicking real membranes.[4] Brain lipid, especially the polar fraction play a key role in the inhibition of prion seeded amyloid formation.[5]

Packaging

5 mL Clear Glass Sealed Ampule (141101P-100mg)
5 mL Clear Glass Sealed Ampule (141101P-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid screening of brain-penetrable antioxidants from natural products by blood-brain barrier specific permeability assay combined with DPPH recognition
Zhu J, et al.
Journal of Pharmaceutical and Biomedical Analysis, 151, 42-48 (2018)
David S Wexler et al.
Journal of biomolecular screening, 10(4), 383-390 (2005-06-21)
Solubility and permeability are intimately linked in drug absorption processes. They have, however, been traditionally assayed separately. To support this linkage, a combined solubility/permeability assay was developed for determining absorption properties of chemical entities. First, solubility is determined at 4
Clearance-dependent underprediction of in vivo intrinsic clearance from human hepatocytes: comparison with permeabilities from artificial membrane (PAMPA) assay, in silico and caco-2 assay, for 65 drugs
Hallifax D, et al.
European Journal of Pharmaceutical Sciences, 45(5), 570-574 (2012)
Laura Campello et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 54(1), 142-159 (2020-02-07)
It is well established that oxidative stress and inflammation are common pathogenic features of retinal degenerative diseases. ITH12674 is a novel compound that induces the transcription factor Nrf2; in so doing, the molecule exhibits anti-inflammatory, and antioxidant properties, and affords
Irene Paterniti et al.
PloS one, 6(1), e15937-e15937 (2011-02-08)
Primary traumatic mechanical injury to the spinal cord (SCI) causes the death of a number of neurons that to date can neither be recovered nor regenerated. During the last years our group has been involved in the design, synthesis and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service