Skip to Content
Merck
All Photos(1)

Key Documents

Z646261

Sigma-Aldrich

Aldrich® drum fittings

vented faucet for 60 L Aldrich drums

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
$154.00

$154.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
5 EA
$154.00

About This Item

UNSPSC Code:
41100000
NACRES:
NB.51

$154.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

diam.

60 mm

Looking for similar products? Visit Product Comparison Guide

General description

Robust PP drum fittings designed to fit Aldrich 5, 10, 20, 30 & 60 L plastic drums, making chemical dispensing from our containers safe and simple.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service