Key Documents
Z567167
Sigma-Aldrich
VerSA-Flow can/drum adapter
57 mm buttress thread to 2 in. NPT
Select a Size
1 EA
$40.50
$40.50
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$40.50
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
$40.50
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
General description
Adapts equipment with standard tapered 2 inch NPT thread such as VerSA-Flow dispensers, pumps, faucets and other hardware to plastic or composite drums with 57 mm Buttress threads.
High density polyethylene construction.
Legal Information
VerSA-Flow is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)