Z566802
Scienceware® Secador® auto-desiccator cabinet
model 3.0, vertical profile, amber, AC/DC input 120 V AC
About This Item
Recommended Products
material
polycarbonate
feature
CSA approved
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F42073-1118
H × W × D
41.7 cm × 34.1 cm × 41.4 cm
door opening H × W
31.2 cm × 22.5 cm
color
amber
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Protecting valuable humidity sensitive items such as reagents or pipettes is easy and totally automatic with Secador Auto-Desiccator Cabinets. Three different size models are available to best match individual storage needs, and can be stacked together to save space.
The 2.0, 3.0 and 4.0 models have internal volumes of 1.1 cu.ft., 1.5 cu. ft., and 1.9 cu.ft. respectively. The cabinets are molded of rugged polycarbonate resistant to staining, crazing and chemical attack.
A patented electronic dehumidification system is designed to reduce the relative humidity inside the cabinet from 75 to 25% and circulate dry air automatically, freeing users form monitoring or changing chemical desiccant.
All contents are easily viewed through a large clear door that seals with positive latches and can be securely closed with tamper evident seals. A hygrometer in the door allows for easy monitoring of relative humidity.
Includes: Perforated, removable shelves.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service