These desiccant beads can be regenerated in an oven at 200°C for 3 hours. The product is specially designed for use with Bel-Art Secador® Auto-Desiccator Cabinet, but may also be used as a desiccant in other systems or applications. Please see the link below to review the Secador® Dessicator Cabinet:
https://www.sigmaaldrich.com/substance/sciencewaresecadorautodesiccatorcabinet1234598765
BAF420740020
SP Bel-Art® Secador® Auto-Desiccator Module Desiccant Silica Replacement Beads
pkg of 26 g
About This Item
Recommended Products
packaging
pkg of 26 g
manufacturer/tradename
Bel-Art F42074-0020
General description
- Silica gel absorbs moisture from the air and is inert, non-toxic and non-corrosive
- Unlike other desiccants, silica gel is non-friable and won’t disintegrate into powder even after repeated regeneration
- Beads contain a color-changing moisture indicator that turns from blue to light pink, indicating that the silica gel is saturated and cannot absorb additional moisture
- When beads turn pink/dark brown, and fail to regain their blue color, it is time to replace the beads with a fresh supply
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
-
I usually dry my beads in an oven. Do these beads require the module to dry them?
1 answer-
Helpful?
-